Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4503190

Sigma-Aldrich

Anti-Prostate Stem Cell Antigen antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PSCA, Prostate stem cell antigen

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$522.00

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$522.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 12 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... PSCA(8000)

Related Categories

General description

Prostate stem cell antigen (PSCA) is a glycosylphosphatidylinositol (GPI)-anchored protein. The gene encoding it is localized on human chromosome 8q24. PSCA is a 123 amino acid protein, belonging to the thymocyte antigen 1 family.

Immunogen

The antiserum was produced against synthesized peptide derived from human Prostate Stem Cell Antigen.

Immunogen Range: 41-90

Biochem/physiol Actions

Prostate stem cell antigen (PSCA) regulates the cell cycle, has a role in signal transduction pathways and mediates c-Myc-stimulated proliferation. It may have a role in prostate cancer. PSCA has been shown to be expressed in various cancers.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Single-nucleotide polymorphisms in PSCA and the risk of breast cancer in a Chinese population.
Wang M, et al.
Oncotarget, 7(19), 27665-27665 (2016)
Amplification and overexpression of PSCA at 8q24 in invasive micropapillary carcinoma of breast.
Meng F, et al.
Breast Cancer Research and Treatment, 166(2), 383-392 (2017)
PSCA regulates IL-6 expression through p38/NF-KB signaling in prostate cancer.
Liu L, et al.
Prostate, 77(14), 1389-1400 (2017)
PSCA promotes prostate cancer proliferation and cell-cycle progression by up-regulating c-Myc.
Li E, et al.
Prostate, 77(16), 1563-1572 (2017)
The influence of PSCA gene variation on its expression and gastric adenocarcinoma susceptibility in the northwest Chinese population.
Zhang W, et al.
International Journal of Molecular Sciences, 16(5), 11648-11658 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service