Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4503088

Sigma-Aldrich

Anti-WEE1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

WEE1hu, Wee1-like protein kinase, kinase WEE1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 71 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... WEE1(7465)

General description

Anti-WEE1 Antibody detects endogenous levels of total WEE1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human WEE1.

Immunogen Range: 19-68

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shuju Zhang et al.
Oncotarget, 7(40), 66087-66099 (2016-09-08)
The anticancer small molecule MLN4924, a Nedd8-activating enzyme (NAE) inhibitor, triggers cell-cycle arrest, apoptosis, and senescence in cancer cells. In this study, we demonstrate that MLN4924 suppresses osteosarcoma cell proliferation by inducing G2/M cell cycle arrest and apoptosis. Our results

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service