Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4503074

Sigma-Aldrich

Anti-VGF, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym(s):

VGF nerve growth factor inducible, neuro-endocrine specific VGF, neurosecretory protein VGF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

rat, human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... VGF(7425)

General description

Anti-VGF Antibody detects endogenous levels of total VGF protein.
The VGF (VGF nerve growth factor inducible) gene is mapped to human chromosome 7q22.1 and codes for a neuropeptide, highly expressed in brain.

Immunogen

The antiserum was produced against synthesized peptide derived from human VGF.

Immunogen Range: 454-503

Biochem/physiol Actions

VGF (VGF nerve growth factor inducible) when induced by neurotrophins such as brain-derived neurotrophic factor, is believed to enhance neuron formation in hippocampus. The gene has an influence on a number of biological processes through its proteolytically digested peptide. Processes include metabolic homeostasis, memory, pain, circadian rhythm, regulation of arginine vasopressin secretion, neurodegeneration and neuropsychiatric disorders. Downregulation of the gene was observed in depression and human bipolar disorder. The encoded growth factor has neuroprotective action on impaired neurons in order to support cell survival.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Neuropeptide VGF Promotes Maturation of Hippocampal Dendrites That Is Reduced by Single Nucleotide Polymorphisms
Behnke J, et al.
International Journal of Molecular Sciences, 18(3), 612-612 (2017)
The Neurotrophin-Inducible Gene Vgf Regulates Hippocampal Function and Behavior through a Brain-Derived Neurotrophic Factor-Dependent Mechanism
Bozdagi O, et al.
The Journal of Neuroscience, 28(39), 9857-9869 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service