Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4503060

Sigma-Aldrich

Anti-UGDH, antibody produced in rabbit

affinity isolated antibody

Synonym(s):

UDP-Glc dehydrogenase, UDP-GlcDH, UDP-glucose 6-dehydrogenase, UDPGDH, UGDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 55 kDa

species reactivity

mouse, human, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... UGDH(7358)

General description

Anti-UGDH Antibody detects endogenous levels of total UGDH protein.
The UGDH (UDP-glucose dehydrogenase) gene is mapped to human chromosome 4p14.

Immunogen

The antiserum was produced against synthesized peptide derived from human UGDH.

Immunogen Range: 391-440

Biochem/physiol Actions

UGDH (UDP-glucose dehydrogenase) catalyzes the dehydrogenation of UDP-glucose to form UDP-glucuronic acid, an important precursor for the glycosaminoglycan chain synthesis of proteoglycans. UGDH is involved in hyaluronan production, steroid elimination and glycosaminoglycan synthesis. UGDH is responsible for the development of circulatory system in different organisms.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Expanded comparative mapping between man and rabbit and detection of a new conserved segment between HSA22 and OCU4
Chantry-Darmon C, et al.
Cytogenetic and genome research, 111(2), 134-139 (2005)
UDP-glucose dehydrogenase modulates proteoglycan synthesis in articular chondrocytes: its possible involvement and regulation in osteoarthritis
Wen Y, et al.
Arthritis Research & Therapy, 16(6), 484-484 (2014)
Annastasia S Hyde et al.
The Journal of biological chemistry, 288(49), 35049-35057 (2013-10-23)
UDP-glucose dehydrogenase (UGDH) provides precursors for steroid elimination, hyaluronan production, and glycosaminoglycan synthesis. The wild-type UGDH enzyme purifies in a hexamer-dimer equilibrium and transiently undergoes dynamic motion that exposes the dimer-dimer interface during catalysis. In the current study we created

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service