Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502469

Sigma-Aldrich

Anti-GPR126 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

GPR126, Probable G-protein coupled receptor 126

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$522.00

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$522.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 136 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunofluorescence: 1:100-1:500

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GPR126(57211)

General description

G protein-coupled receptor 126 (GPR126) belongs to the adhesion-G protein-coupled receptor (GPCR) family. It is an endothelial cell-enriched gene. The gene is located on human chromosome 6q24.2.
The GPR126 (adhesion G protein-coupled receptor G6) gene codes for a protein that contains an extracellular domain, a seven transmembrane domain at the N-terminal and an intracellular domain in its C-terminal.

Immunogen

The antiserum was produced against synthesized peptide derived from human GPR126.

Immunogen Range: 1091-1140

Biochem/physiol Actions

GPR126 (adhesion G protein-coupled receptor G6) controls the homeostasis of periodontal ligament tissue. GPR126 is responsible for axon myelination of peripheral neurons in human, mice and fish. This gene mediates endothelial cell proliferation and migration, and tube formation, thereby contributing to angiogenesis, which is necessary for overall growth, wound healing and tumor progression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A Genetic Variant in GPR126 Causing a Decreased Inclusion of Exon 6 Is Associated with Cartilage Development in Adolescent Idiopathic Scoliosis Population
Xu E, et al.
BioMed Research International, 2019 (2019)
Multiethnic GWAS reveals polygenic architecture of earlobe attachment
Shaffer JR, et al.
American Journal of Human Genetics, 101(6), 913-924 (2017)
Gianina Ravenscroft et al.
American journal of human genetics, 96(6), 955-961 (2015-05-26)
Arthrogryposis multiplex congenita is defined by the presence of contractures across two or more major joints and results from reduced or absent fetal movement. Here, we present three consanguineous families affected by lethal arthrogryposis multiplex congenita. By whole-exome or targeted
Identification of Novel Smoothened Ligands Using Structure-Based Docking.
Lacroix C
PLoS ONE, 11(8), 1-20 (2016)
Hengxiang Cui et al.
The Journal of biological chemistry, 289(50), 34871-34885 (2014-09-14)
Angiogenesis, the formation of new blood vessels from pre-existing ones, is essential for development, wound healing, and tumor progression. The VEGF pathway plays irreplaceable roles during angiogenesis, but how other signals cross-talk with and modulate VEGF cascades is not clearly

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service