Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502328

Sigma-Aldrich

Anti-ZNF280C antibody produced in rabbit

affinity isolated antibody

Synonym(s):

SUHW3, Suppressor of hairy wing homolog 3, ZNF280C, Zinc finger protein 280C

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 83 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF280C(55609)

General description

Anti-ZNF280C Antibody detects endogenous levels of total ZNF280C protein.
Zinc-fingers (ZnFs) are ubiquitously present in higher eukaryotes. ZNF280C (zinc finger protein 280C) is located on human chromosome X.

Immunogen

The antiserum was produced against synthesized peptide derived from human ZNF280C.

Immunogen Range: 251-300

Biochem/physiol Actions

Zinc-fingers (ZnFs) serves as sequence-specific DNA-binding motifs. It also plays an important role in recognition of RNA and other proteins.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sticky fingers: zinc-fingers as protein-recognition motifs.
Gamsjaeger R, et al.
Trends in Biochemical Sciences, 32(2), 63-70 (2007)
Derivation of new human embryonic stem cell lines reveals rapid epigenetic progression in vitro that can be prevented by chemical modification of chromatin.
Diaz Perez SV, et al.
Human Molecular Genetics, 21(4), 751-764 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service