Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4502044

Sigma-Aldrich

Anti-ZNF295 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KIAA1227, ZBTB21, ZNF295, Zinc finger and BTB domain-containing protein 21, Zinc finger protein 295

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$392.00

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$392.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 118 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZNF295(49854)

General description

Anti-ZNF295 Antibody detects endogenous levels of total ZNF295 protein.
The gene ZBTB21 (zinc finger and BTB domain containing 21) is mapped to human chromosome 21q22.3. The protein contains a BTB (broad-complex, tramtrack and bric a brac)/POZ (poxvirus and zinc finger) domain and multiple kruppel (C2H2) type zinc finger domains. ZBTB21 is part of the POK (POZ and krüppel) transcription factor family of proteins. It is widely expressed in human tissues.

Immunogen

The antiserum was produced against synthesized peptide derived from human ZNF295.

Immunogen Range: 731-780

Biochem/physiol Actions

ZBTB21 (zinc finger and BTB domain containing 21) is a transcription repressor. It interacts with ZFP161 (zinc finger protein 161) and might be associated with the bi-directional control of gene expression.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Reza Sailani et al.
Genome research, 23(9), 1410-1421 (2013-06-21)
Congenital heart defect (CHD) occurs in 40% of Down syndrome (DS) cases. While carrying three copies of chromosome 21 increases the risk for CHD, trisomy 21 itself is not sufficient to cause CHD. Thus, additional genetic variation and/or environmental factors
Jun Wang et al.
Biochemical and biophysical research communications, 327(2), 615-627 (2005-01-05)
Human ZNF295 protein harbors a BTB/POZ domain and multiple krüppel (C(2)H(2)) type zinc finger domains, and thus belongs to a family of POK (POZ and krüppel) transcription factor. We have identified many transcript variants generated by the alternative splicing in

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service