Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501906

Sigma-Aldrich

Anti-MMP17 (Cleaved-Gln129) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

MMP-17, MT-MMP 4, MT4-MMP, MT4MMP, matrix metalloproteinase-17

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$392.00

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$392.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 53 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

proteolytically cleaved (Gln129)

Gene Information

human ... MMP17(4326)

Related Categories

General description

Anti-MMP17 (Cleaved-Gln129) Antibody detects endogenous levels of fragment of activated MMP17 (Cleaved-Gln129) protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human MMP17.

Immunogen Range: 110-159

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T J Kaitu'u-Lino et al.
Placenta, 34(2), 168-173 (2012-12-25)
Pre-eclampsia is a serious complication of pregnancy, characterized by severe endothelial dysfunction resulting in hypertension, proteinuria and maternal end-organ damage. Soluble endoglin is an anti-angiogenic factor released from placenta that has been linked to severe pre-eclampsia. We recently reported MMP-14

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service