Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501864

Sigma-Aldrich

Anti-MAP3K4 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

KIAA0213, M3K4, MAP three kinase 1, MAP3K4, MAPK/ERK kinase kinase 4

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$392.00

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$392.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 181 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:5000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAP3K4(4216)

General description

Anti-MAP3K4 Antibody detects endogenous levels of total MAP3K4 protein.
MAP3K4 (mitogen-activated protein kinase kinase kinase 4) gene encodes a component of protein kinase signal transduction cascade. It is also termed as MTK1 (MAP3 kinase 1). MAP3K4 contains a protein kinase catalytic domain at the C terminus and a regulatory/autoinhibitory domain at the N-terminus. It is located on human chromosome 6q26.

Immunogen

The antiserum was produced against synthesized peptide derived from human MAP3K4.

Immunogen Range: 1281-1330

Biochem/physiol Actions

MAP3K4 (mitogen-activated protein kinase kinase kinase 4) phosphorylates and activates a specific MAPK kinase (MAPKK) in the cascade through phosphorylation of conserved threonine and/or serine residues. GADD45 (growth arrest and DNA-damage-inducible proteins) proteins interact with and free the autoinhibitory domain from the kinase domain, thus activating MAP3K4. It mediates the activation of p38 MAPK pathway and the JNK (c-Jun N-terminal kinase) pathway induced by environmental stresses.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Comparison between osteoblasts derived from human dental pulp stem cells and osteosarcoma cell lines
Palmieri A, et al.
Cell Biology International, 32(7), 733-738 (2008)
Hiroaki Mita et al.
Molecular and cellular biology, 22(13), 4544-4555 (2002-06-08)
A variety of cellular stresses activate the stress-responsive mitogen-activated protein (MAP) kinases p38 and JNK. In this study, we studied the activation mechanism of a human MAP kinase kinase kinase, MTK1 (also known as MEKK4), which mediates activation of both
M Takekawa et al.
The EMBO journal, 16(16), 4973-4982 (1997-08-15)
A human homolog of the yeast Ssk2 and Ssk22 mitogen-activated protein kinase kinase kinases (MAPKKK) was cloned by functional complementation of the osmosensitivity of the yeast ssk2delta ssk22delta sho1delta triple mutant. This kinase, termed MTK1 (MAP Three Kinase 1), is

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service