Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4501600

Sigma-Aldrich

Anti-JAK2 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

JAK-2, JAK2, Janus kinase 2, kinase Jak2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 130 kDa

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... JAK2(3717)

General description

Anti-JAK2 Antibody detects endogenous levels of total JAK2 protein.
JAK2 (janus kinase 2) encodes a tyrosine kinase. It belongs to the Jak family of proteins. The JAK2 gene is located on human chromosome 9p24.

Immunogen

The antiserum was produced against synthesized peptide derived from human JAK2.

Immunogen Range: 981-1030

Biochem/physiol Actions

JAK2 (janus kinase 2) plays an important role in the activities of cytokine receptors. Overexpression of JAK2 results in neuroblastoma. JAK2 mutations promote primary resistance to PD-1 (anti-programmed death protein 1) blockade therapy.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hodgkin's lymphoma cell lines are characterized by frequent aberrations on chromosomes 2p and 9p including REL and JAK2.
Stefan J, et al.
Cancer Genetics, 103(4), 489-495 (2002)
Jak2 is essential for signaling through a variety of cytokine receptors
Parganas E, et al.
Cell, 93(3), 385-395 (1998)
Ekaterina A Polyakova et al.
Heliyon, 7(11), e08491-e08491 (2021-12-14)
Hyperleptinemia potentiates the effects of many atherogenic factors, such as inflammation, platelet aggregation, migration, hypertrophy, proliferation of vascular smooth muscle cells, and endothelial cell dysfunction. The present study analysed the effects of long-term hyperleptinemia in an in vivo myocardial ischemia-reperfusion
Cancer-Associated Fibroblasts Share Characteristics and Protumorigenic Activity with Mesenchymal Stromal Cells.
Borriello L
Cancer Research, 77(18), 5142-5157 (2017)
Primary Resistance to PD-1 Blockade Mediated by JAK1/2 Mutations.
Shin DS
Cancer Discovery, 7(2), 188-201 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service