Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4501584

Sigma-Aldrich

Anti-CD18 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

95 β, CD18, cell surface adhesion glycoproteins LFA- 1/CR3/p150, complement receptor C3 β, integrin β-2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 84 kDa

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ITGB2(3689)

General description

Anti-CD18 Antibody detects endogenous levels of total CD18 protein.
CD18/ ITGB2 (integrin subunit β2) is also termed as Mac-1 or complement receptor 3. The ITGB2 gene is mapped to human chromosome 21q22.3. It is mainly expressed in blood. Anti-CD18 Antibody detects endogenous levels of total CD18 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD18/ITGB2.

Immunogen Range: 720-769

Biochem/physiol Actions

CD18/ ITGB2 (integrin subunit β 2) plays an important role in the progression and function of T-cells. Mutations in ITGB2 results in leukocyte adhesion deficiency type 1, which has an established role in T-cell development and function. ITGB2/CD18 is associated with chronic colitis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Use of degenerate oligonucleotide primed PCR (DOP-PCR) for the genotyping of low-concentration DNA samples.
Barbaux S, et al.
Journal of Molecular Medicine, 79(5-6), 329-332 (2001)
Altered levels of soluble CD18 may associate immune mechanisms with outcome in sepsis.
Kragstrup TW, et al.
Clinical and Experimental Immunology, 190(2), 258-267 (2017)
Evaluating a CLL susceptibility variant in ITGB2 in families with multiple subtypes of hematological malignancies.
Nicholas BB, et al.
Blood, 130(1), 86?88-86?88 (2017)
The ITGB2 immunomodulatory gene (CD18), enterocolitis, and Hirschsprung's disease.
Moore SW, et al.
Journal of Pediatric Surgery, 43(8), 1439-1444 (2008)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service