Anti-IRS-1 Antibody detects endogenous levels of total IRS-1 protein.
Insulin receptor substrate 1 (IRS1) belongs to the IRS family. It is mapped to human chromosome 2q36.
Immunogen
The antiserum was produced against synthesized peptide derived from human IRS-1.
Immunogen Range: 289-338
Biochem/physiol Actions
Insulin receptor substrate 1 (IRS1) participates in the insulin signaling pathway. It is associated with type 2 diabetes (T2D). IRS1 regulates glucose homeostasis, body growth and peripheral insulin action.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
The type 2 diabetes and insulin-resistance locus near IRS1 is a determinant of HDL cholesterol and triglycerides levels among diabetic subjects.
Sharma R, et al.
Atherosclerosis, 216(1), 157-160 (2011)
Association between IRS1 Gene Polymorphism and Autism Spectrum Disorder: A Pilot Case-Control Study in Korean Males.
Park HJ, et al.
International Journal of Molecular Sciences, 17(8), 1227-1227 (2016)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.