Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4501527

Sigma-Aldrich

Anti-IGFBP-3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

IBP-3, IBP3, IGF-binding protein 3, IGFBP3, Insulin-like growth factor binding protein 3 precursor

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 31 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IGFBP3(3486)

General description

Insulin-like growth factor-binding protein 3 (IGFBP3), a multifunctional protein belongs to the insulin-like growth factor binding proteins (IGFBPs) superfamily. The IGFBP3 gene is located on human chromosome 7p12. Anti-IGFBP-3 antibody detects endogenous levels of total IGFBP-3 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human IGFBP-3.

Immunogen Range: 151-200

Biochem/physiol Actions

Insulin-like growth factor-binding protein 3 (IGFBP3) can stimulate cell growth and apoptosis. It participates in DNA repair. Patients with rheumatoid arthritis show an increased level of IGFBP3 in the serum and synovial fluid. It is involved in the proliferation of stress-induced senescence in human endometrium-derived mesenchymal stem cells (MESCs). IGFBP3 can suppress insulin-like growth factor-1 (IGF-1) signaling and modulate lung carcinogenesis and cell proliferation.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Eggermann et al.
Annales de genetique, 42(2), 117-121 (1999-08-06)
Maternal uniparental disomy (UPD) of chromosome 7 has recently been reported in about 10% of Silver-Russell (SRS) patients. It can therefore be concluded that at least one gene on chromosome 7 is imprinted and mutations in this gene/these genes might
Roman E Ushakov et al.
Biochemical and biophysical research communications, 531(2), 133-139 (2020-08-13)
Insulin-like growth factor binding protein 3 (IGFBP3) is a multifunctional protein, able either to stimulate the cell growth or to promote apoptosis. In particular, IGFBP3 plays significant role in propagation of stress-induced senescence in human endometrium-derived mesenchymal stem cells (MESCs)
Chia-Chia Chao et al.
Life sciences, 265, 118758-118758 (2020-11-15)
Insulin-like growth factor (IGF) signaling has been documented in several human malignancies and is thought to contribute to cellular differentiation and migration, as well as malignant progression. A major binding molecule of IGF, IGF-binding protein 3 (IGFBP-3), regulates multiple IGF

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service