Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4501207

Sigma-Aldrich

Anti-SESN1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

SESN1, Sestrin-1, p53-regulated protein PA26

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 56 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
immunohistochemistry: 1:50-1:100
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SESN1(27244)

General description

Anti-SESN1 Antibody detects endogenous levels of total SESN1 protein.
Sestrins are stress-inducible proteins. The SESN1 (sestrin 1) gene is located on human chromosome 6q21.

Immunogen

The antiserum was produced against synthesized peptide derived from human SESN1.

Immunogen Range: 271-320

Biochem/physiol Actions

Knockdown of SESN1 (sestrin 1) blocks cell cycle progression. Sestrin 1 acts as an antioxidant and as inhibitor of target of rapamycin complex 1. It is considered as a p53 target gene and it helps to connect between genotoxic stress, p53 and the mTOR signaling pathway.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

p53 target genes sestrin1 and sestrin2 connect genotoxic stress and mTOR signaling
Budanov AV and Karin M
Cell, 134(3), 451-460 (2008)
A study of recombinant human sestrin 1 and sestrin 2 proteins produced in a prokaryotic system
Rai N, et al.
Molekuliarnaia Biologiia, 51(3), 416-425 (2017)
Suppressed expression of non-DSB repair genes inhibits gamma-radiation-induced cytogenetic repair and cell cycle arrest
Zhang Y, et al.
DNA Repair, 7, 1835-1845 (2008)
Sestrins at the crossroad between stress and aging
Lee JH, et al.
Aging (Albany. NY.), 2 (2010)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service