Anti-GABRG1 Antibody detects endogenous levels of total GABRG1 protein. GABRG1 (γ-aminobutyric acid type A receptor γ1 subunit) is a member of the ligand-gated ionic channel family. It is mapped to human chromosome 4p.
Immunogen
The antiserum was produced against synthesized peptide derived from human GABRG1.
Immunogen Range: 11-60
Biochem/physiol Actions
Mutation in GABRG1 (γ-aminobutyric acid type A receptor γ1 subunit) leads to alcohol dependence.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.
Genetic association of recovery from eating disorders: the role of GABA receptor SNPs.
Bloss CS, et al.
Neuropsychopharmacology, 36(11), 2222-2222 (2011)
GABRG1 and GABRA2 variation associated with alcohol dependence in African Americans.
Ittiwut C, et al.
Alcoholism, Clinical and Experimental Research, 36(4), 588-593 (2012)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.