Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4501000

Sigma-Aldrich

Anti-FOSB antibody produced in rabbit

affinity isolated antibody

Synonym(s):

G0/G1 switch regulatory protein 3, G0S3, Protein fosB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... FOSB(2354)

General description

Anti-FOSB Antibody detects endogenous levels of total FOSB protein.

FosB (proto-oncogene, AP-1 transcription factor subunit) is transcription factor that belongs to Fos family of transcription factors. It is a ~50 kDa protein. FosB gene codes for δFosB (δ FosB). It is mapped to human chromosome 19q13.

Immunogen

The antiserum was produced against synthesized peptide derived from human FosB.

Immunogen Range: 12-61

Biochem/physiol Actions

FosB (proto-oncogene, AP-1 transcription factor subunit) acts as a diagnostic marker for pseudomyogenic hemangioendothelioma. δFosB controls multiplication, differentiation and death in epithelial cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High-throughput tissue microarray analysis of cyclin E gene amplification and overexpression in urinary bladder cancer
Richter J, et al.
The American Journal of Pathology, 157(3), 787-794 (2000)
?FosB: a transcriptional regulator of stress and antidepressant responses
Nestler EJ
European Journal of Pharmacology, 753, 66-72 (2015)
FOSB is a Useful Diagnostic Marker for Pseudomyogenic Hemangioendothelioma.
Hung YP, Fletcher CD, Hornick JL.
American Journal of Surgical Pathology, 41(5), 596-606 (2017)
Regulatory effects of ?FosB on proliferation and apoptosis of MCF-7 breast cancer cells
Li H, et al.
Tumour Biology : the Journal of the International Society For Oncodevelopmental Biology and Medicine, 37(5), 6053-6063 (2016)
Differential Expression of FosB Proteins and Potential Target Genes in Select Brain Regions of Addiction and Depression Patients
Gajewski PA, et al.
PLoS ONE, 11(8), e0160355-e0160355 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service