Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500988

Sigma-Aldrich

Anti-PHF3 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PHD finger protein 3

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$522.00

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$522.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 229 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... PHF3(23469)

General description

Anti-PHF3 Antibody detects endogenous levels of total PHF3 protein.
The gene PHF3 (PHD finger protein 3) is mapped to human chromosome 6. It is widely expressed in human tissues.

Immunogen

The antiserum was produced against synthesized peptide derived from human PHF3.

Immunogen Range: 1990-2039

Biochem/physiol Actions

The gene region PTP4A1-PHF3-EYS (protein tyrosine phosphatase type 4A member 1 - PHD finger protein 3 - eyes shut homolog) is associated with alcohol dependence.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Association of rare PTP4A1-PHF3-EYS variants with alcohol dependence.
Lingjun Zuo et al.
Journal of human genetics, 58(3), 178-179 (2013-01-18)
Lingjun Zuo et al.
PloS one, 6(11), e26726-e26726 (2011-11-19)
Several genome-wide association studies (GWASs) reported tens of risk genes for alcohol dependence, but most of them have not been replicated or confirmed by functional studies. The present study used a GWAS to search for novel, functional and replicable risk
Lingjun Zuo et al.
The American journal on addictions, 23(4), 411-414 (2014-06-26)
We previously reported a risk genomic region (ie, PTP4A1-PHF3-EYS) for alcohol dependence in a genome-wide association study (GWAS). We also reported a rare variant constellation across this region that was significantly associated with alcohol dependence. In the present study, we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service