Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500944

Sigma-Aldrich

Anti-AKR1B1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

AR, Aldehyde reductase, Aldose reductase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 35 kDa

species reactivity

rat, human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... AKR1B1(231)

General description

Aldo-keto reductase family 1, member B1 (AKR1B1) is part of the aldo/keto reductase superfamily. The gene encoding it is localized on human chromosome 7. Anti-AKR1B1 Antibody detects endogenous levels of total AKR1B1 protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human AKR1B1.

Immunogen Range: 241-290

Biochem/physiol Actions

Aldo-keto reductase family 1, member B1 (AKR1B1) takes part in the reduction of aldehydes. It can be induced by proteasome inhibitors in human colon cancer cell lines HT-29 and SW-480.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Variants in the gene encoding aldose reductase (AKR1B1) and diabetic nephropathy in American Indians.
Wolford JK
Diabetic Medicine : a Journal of the British Diabetic Association, 23(4), 367-376 (2006)
Bettina Ebert et al.
Chemico-biological interactions, 191(1-3), 239-249 (2011-01-11)
Aldo-keto reductases (AKRs) play central roles in the reductive metabolism of endogenous signaling molecules and in the detoxification of xenobiotics. AKRC1-1C3, AKR1B1 and AKR1B10 have been shown to be regulated via nuclear factor-erythroid 2 related factor 2 (Nrf2), a transcription
Pushplata Prasad et al.
BMC medical genetics, 11, 52-52 (2010-04-01)
To determine association of nine single nucleotide polymorphisms (SNPs) in ADP ribosyltransferase-1 (ADPRT1), aldo-keto reductase family 1 member B1 (AKR1B1), receptor for advanced glycation end-products (RAGE), glutamine:fructose-6-phosphate amidotransferase-2 (GFPT2), and plasminogen activator inhibitor-1 (PAI-1) genes with chronic renal insufficiency (CRI)
Liping Zhang et al.
FEBS letters, 587(22), 3681-3686 (2013-10-09)
The antineoplastic target aldo-keto reductase family member 1B10 (AKR1B10) and the critical polyol pathway enzyme aldose reductase (AKR1B1) share high structural similarity. Crystal structures reported here reveal a surprising Trp112 native conformation stabilized by a specific Gln114-centered hydrogen bond network

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service