Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500770

Sigma-Aldrich

Anti-ADCK5 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

ADCK5, Uncharacterized aarF domain-containing protein kinase 5

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$392.00

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.


Select a Size

Change View
100 μG
$392.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$392.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

Notify Me

Get notified when this item is ready to ship via email.

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 65 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:20000
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ADCK5(203054)

General description

Anti-ADCK5 Antibody detects endogenous levels of total ADCK5 protein.
The gene ADCK5 (aarF domain containing kinase 5) is mapped to human chromosome 8q24.3. It is a putative atypical kinase. ADCK5 might be involved in the coenzyme Q biosynthetic pathway.

Immunogen

The antiserum was produced against synthesized peptide derived from human ADCK5.

Immunogen Range: 151-200

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Coenzyme Q biosynthesis in health and disease.
Acosta MJ, et al.
Biochimica et Biophysica Acta, 1857, 1079-1085 (2016)
Genetic bases and clinical manifestations of coenzyme Q10 (CoQ 10) deficiency.
Desbats MA, et al.
Journal of inherited metabolic disease, 38, 145-156 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service