Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500696

Sigma-Aldrich

Anti-EFNA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

B61, ECKLG, EFL1, EPH-related receptor tyrosine kinase ligand 1, ephrin-A1

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$522.00

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)


Select a Size

Change View
100 μG
$522.00

About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

$522.00


Usually ships in 5 business days. (Orders outside of US, please allow an additional 1-2 weeks for delivery)

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 23 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:10000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... EFNA1(1942)

General description

EphrinA1 (EFNA1), also known as angiogenesis factor, is encoded by the gene mapped to human chromosome 1q21-22. The encoded protein belongs to subclass A of ephrin family. EFNA1 is localized to cell membrane via a glycosylphosphatidylinositol (GPI) moiety. EFNA1 ligand preferentially binds to the receptor tyrosine kinase, Eph receptor 2 (EphA2).

Immunogen

The antiserum was produced against synthesized peptide derived from human EFNA1.

Immunogen Range: 66-115

Application

Anti-EFNA1 antibody produced in rabbit has been used in western blot analysis.[1]

Biochem/physiol Actions

EphrinA1 (EFNA1) and its primary receptor, Eph receptor 2 (EphA2) are associated with various cancers, such as urinary bladder carcinoma, breast cancer gastric cancer, glioma and malignant mesothelioma. Expression pattern of EFNA1 can be used as a potential prognostic marker in colorectal cancer (CRC) patients and other human malignancies. Experimental studies state that, EphrinA1 (EFNA1) can be involved in both tumor promotion and inhibition, based on whether ephrin-A1 is bound to the receptor in soluble or membrane bound form, respectively. Activation of EphA2, induces indirect growth signal by overcoming ‘contact inhibition′.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The genes encoding the eph-related receptor tyrosine kinase ligands LERK-1 (EPLG1, Epl1), LERK-3 (EPLG3, Epl3), and LERK-4 (EPLG4, Epl4) are clustered on human chromosome 1 and mouse chromosome 3.
Cerretti DP, et al.
Genomics, 33(2), 277-282 (1996)
Lyka Potla et al.
Cancer letters, 175(2), 187-195 (2001-12-14)
Ephrin A1 (EFNA1) is a GPI-anchored ligand that preferentially binds to the receptor tyrosine kinase, EphA2. EphA2 is over-expressed in malignant melanocytes and in prostate carcinoma cells. Whether activation of EphA2 by EFNA1 is involved in aberrant growth or differentiation
T Tawadros et al.
British journal of cancer, 107(10), 1737-1744 (2012-10-06)
High intake of omega-6 polyunsaturated fatty acids (PUFA) has been associated with clinical progression in prostate cancer (CaP). This study investigates the signalling mechanism by which the omega-6 PUFA arachidonic acid (AA) induces prostatic cellular migration to bone marrow stroma.
Hirofumi Yamamoto et al.
International journal of oncology, 42(2), 549-555 (2012-12-22)
We previously studied hypoxic tumor cells from hepatic metastases of colorectal cancer (CRC) and determined several potential prognostic factors, including expression
Sho Kohara et al.
Biomedicines, 10(12) (2022-12-24)
Kupffer cells are maintained via self-renewal in specific microenvironmental niches, primarily the liver sinusoidal endothelial cells (LSECs). In this study, we propagated tissue-resident macrophages (Mø) from mouse liver using mixed culture with hepatic fibroblastic cells. Propagated liver Mø express Id3

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service