Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500626

Sigma-Aldrich

Anti-DCT antibody produced in rabbit

affinity isolated antibody

Synonym(s):

DCT, DT, L-dopachrome δ-isomerase, L-dopachrome tautomerase, Tyrosinase-related protein 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μG
$522.00

$522.00


Check Cart for Availability


Select a Size

Change View
100 μG
$522.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$522.00


Check Cart for Availability

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 59 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TYRP2(1638)

General description

Anti-DCT Antibody detects endogenous levels of total DCT protein.
Dopachrome tautomerase (DCT) is also termed as tyrosinase-related protein 2 (TYRP2). It is present in melanocytes, RPE (retinal pigment epithelium) cells and HaCaTs (basal keratinocytes). It is located on human chromosome 13q32.

Immunogen

The antiserum was produced against synthesized peptide derived from human DCT.

Immunogen Range: 51-100

Biochem/physiol Actions

Dopachrome tautomerase (DCT) participates in the production of melanin in melanosomes and also in the detoxification of the melanin precursors. It restricts the destructions induced by chemotherapeutic agents like X-ray or UVB (ultraviolet B) and gamma irradiation in melanoma cells and reduces the susceptibility of cell to oxidative stress.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The Role of DCT in HPV16 Infection of HaCaTs
Aksoy P and Meneses PI
PLoS ONE (2017)
Finding genes that underlie physical traits of forensic interest using genetic tools
Pulker H, et al.
Forensic Science International. Genetics, 1(2), 100-104 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service