Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500177

Sigma-Aldrich

Anti-GA45G antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Cytokine-responsive protein CR6, DDIT-2, DNA-damage-inducible transcript 2, GADD45G, Growth arrest and DNA-damage-inducible protein GADD45 γ

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 17 kDa

species reactivity

human, mouse, rat

concentration

~1 mg/mL

technique(s)

ELISA: 1:1000
immunofluorescence: 1:100-1:500
immunohistochemistry: 1:50-1:100

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GADD45G(10912)

General description

The GADD45G (growth arrest and DNA damage-inducible gamma) gene is mapped to human chromosome 9q22.2. This gene is highly conserved along evolution. It is found to be localized in various tissues, both in adult and fetus. Anti-GA45G antibody detects endogenous levels of total GA45G protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human GA45G.

Immunogen Range: 101-150

Application

Anti-GA45G antibody produced in rabbit has been used in western blot analysis.

Biochem/physiol Actions

Growth arrest DNA damage-inducible 45 (GADD45G) has its key role in regulating DNA repair, cell cycle and apoptosis. It regulates cellular action in response to physical and environmental stress. GADD45G is considered as a stress-responsive protein. It controls the functions of proliferating cell nuclear antigen (PCNA), p21, Cdk1 (cyclin-dependent kinase 1), cdc2 (cell division cycle protein 2)/cyclin B1, p38 and c-Jun N-terminal kinase (JNK). GADD45G is associated with the disease progression of diffuse large B-cell lymphoma (DLBL). It is known to be hypermethylated in DLBL. Lowered GADD45G gene expression is observed in solid tumors.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

SIP1 is a downstream effector of GADD45G in senescence induction and growth inhibition of liver tumor cells
Xu G, et al.
Oncotarget, 6(32), 33636-33636 (2015)
Possible Role of GADD45gamma Methylation in Diffuse Large B-Cell Lymphoma: Does It Affect the Progression and Tissue Involvement?
Barics IC
Turkish journal of haematology : official journal of Turkish Society of Haematology, 32(4), 295-295 (2015)
Nasopharyngeal carcinoma: molecular pathogenesis and therapeutic developments
Tao Q and Chan ATC
Expert Reviews in Molecular Medicine, 9(12), 1-24 (2007)
Alexandre Lucas et al.
Cardiovascular research, 108(2), 254-267 (2015-09-16)
Post-infarction remodelling is accompanied and influenced by perturbations in mitogen-activated protein kinase (MAPK) signalling. The growth arrest and DNA-damage-inducible 45 (Gadd45) proteins are small acidic proteins involved in DNA repair and modulation of MAPK activity. Little is known about the
Gadd45 gamma regulates cardiomyocyte death and post-myocardial infarction left ventricular remodelling
Lucas A, et al.
Cardiovascular Research, 108(2), 254-267 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service