Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4500169

Sigma-Aldrich

Anti-CD3EAP antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-sense to ERCC-1 protein, ASE-1, CD3-ε-associated protein, DNA-directed RNA polymerase I subunit RPA34, RNA polymerase I-associated factor PAF49

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 54 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
western blot: 1:500-1:1000

NCBI accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... RPA34(10849)

General description

The CD3EAP (CD3e molecule, epsilon-associated protein) gene is mapped to human chromosome 19q13.3. It encodes a DNA-dependent RNA polymerase I subunit RPA34 (replication protein A34). This protein is localized to the nucleolus, in the fibrillar centers during interphase and the nucleolus organizer regions of the chromosomes during cell division. Anti-CD3EAP antibody detects endogenous levels of total CD3EAP protein.

Immunogen

The antiserum was produced against synthesized peptide derived from human CD3EAP.

Immunogen Range: 441-490

Biochem/physiol Actions

The CD3EAP (CD3e molecule, epsilon-associated protein) gene participates in the process of programmed cell death and ribosomal RNA transcription. CD3EAP expression is implicated in the pathogenesis of non-small cell lung cancer (NSCLC).

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

19p13. 3-GADD45B common variants and 19q13. 3-PPP1R13L and 19q13. 3-CD3EAP in lung cancer risk among Chinese
Hou W, et al.
Chemico-Biological Interactions, 277, 74-78 (2017)
Effects of PPP1R13L and CD3EAP variants on lung cancer susceptibility among nonsmoking Chinese women
Yin J, et al.
Gene, 524(2), 228-231 (2013)
Different splicing isoforms of ERCC1 affect the expression of its overlapping genes CD3EAP and PPP1R13L, and indicate a potential application in non-small cell lung cancer treatment
Zhang G, et al.
International Journal of Oncology, 52(6), 2155-2165 (2018)
Association of chromosome 19 to lung cancer genotypes and phenotypes.
Wang X
Cancer Metastasis Reviews, 34(2), 217-226 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service