Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4500044

Sigma-Aldrich

Anti-CDH17 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

CAD17, CADH, Intestinal peptide-associated transporter HPT-1, LI-cad, LI-cadherin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 92 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

ELISA: 1:40000
immunofluorescence: 1:100-1:500
western blot: 1:500-1:1000

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDH17(1015)

General description

Cadherin-17 is a liver-intestine cadherin protein encoded by the CDH17 gene in humans and belongs to unique member of cadherin superfamily. CDH17 is a non-classical cadherin having seven cadherin domains and a transmembrane domain but lacking the conserved cytoplasmic domain of classical cadherins. Cadherins are cellular adhesion molecules that are crucial for intracellular adhesion. These are considered as potential tumor suppressor genes and any alterations in them may lead to invasion and metastasis of tumor cells. The gene CDH17 is mapped to human chromosome 8q22.1 and contains 18 exons.

Immunogen

The antiserum was produced against synthesized peptide derived from human CDH17.

Immunogen Range: 341-390

Biochem/physiol Actions

The protein encoded by the gene CDH17 (Cadherin-17), also referred to as LI-cadherin, functions in cell adhesion without being dependent on cytoplasmic components, such as catenins or the actin cytoskeleton. Lower expression of this protein due to single nucleotide polymorphisms has been associated with tumor progression and lymph node metastasis of human colorectal carcinoma. It is found to regulate the signaling function of α2β1 integrin in cell adhesion and proliferation in liver metastatic cancer cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Kremmidiotis et al.
Genomics, 49(3), 467-471 (1998-06-06)
This report presents the chromosomal localization of cadherin genes. Cadherins are cellular adhesion molecules. Since disturbance of intracellular adhesion is important for invasion and metastasis of tumor cells, cadherins are considered prime candidates for tumor suppressor genes. A variety of
R A Bartolomé et al.
Oncogene, 33(13), 1658-1669 (2013-04-23)
Liver metastasis is the major cause of death associated to colorectal cancer. Cadherin-17 (CDH17) is a non-classical, seven domain, cadherin lacking the conserved cytoplasmic domain of classical cadherins. CDH17 was overexpressed in highly metastatic human KM12SM and present in many
Jin Wang et al.
Cancer biology & therapy, 14(3), 262-270 (2013-01-10)
Cadherin-17 (CDH17), as a structurally unique member of the cadherin superfamily, has been identified to predict a poor prognosis for gastric cancer (GC). Our previous study demonstrated the positive correlation between CDH17 and lymph node micrometastasis in GC. We sought
Single nucleotide polymorphisms in the CDH17 gene of colorectal carcinoma.
Chen RY
World Journal of Gastroenterology, 18, 7251-7261 (2012)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service