Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4301448

Sigma-Aldrich

Anti-phospho-VE-Cadherin (pTyr731) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

7B4 antigen, CD144 antigen, CDH5, VE-cadherin, Vascular endothelial cadherin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

130 kDa

species reactivity

rat, mouse, human

concentration

1.0 mg/mL

technique(s)

western blot: 1:500-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001786.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

phosphorylation (pTyr731)

Gene Information

human ... CDH5(1003)

General description

Vascular endothelial cadherin (VE-cadherin) or CDH5 gene is mapped to human chromosome 16q21. It comprises extracellular cadherin (EC) domains that take up a strand-swapped dimers structure.

Specificity

The antibody detects endogenous level of VE-Cadherin only when phosphorylated at tyrosine 731.

Immunogen

Peptide sequence around phosphorylation site of Tyrosine731 (H-I-Y(p)-G-Y) derived from Human VE-Cadherin.

Application

Anti-phospho-VE-Cadherin (pTyr731) antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Vascular endothelial cadherin (VE-cadherin) plays a key role in maintaining membrane integrity and vascular endothelium. It mediates endothelial cell-cell adhesion in particular at the adherens junctions and participates in leukocyte trafficking. VE-cadherin is also needed for embryonic angiogenesis.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julia Brasch et al.
Journal of molecular biology, 408(1), 57-73 (2011-01-29)
Vascular endothelial cadherin (VE-cadherin), a divergent member of the type II classical cadherin family of cell adhesion proteins, mediates homophilic adhesion in the vascular endothelium. Previous investigations with a bacterially produced protein suggested that VE-cadherin forms cell surface trimers that
Mintu Pal et al.
Experimental cell research, 368(2), 137-146 (2018-04-21)
The major hallmarks of Epithelial-Mesenchymal Transition (EMT) is the loss of epithelial cell polarity and loss of expression of the cell- cell adhesion molecule like E-cadherin and acquired mesenchymal cells marker called N-Cadherin. This phenotypical changes of E-M plasticity of
Lihua Ying et al.
American journal of physiology. Lung cellular and molecular physiology, 315(1), L66-L77 (2018-03-31)
Compromised pulmonary endothelial cell (PEC) barrier function characterizes acute respiratory distress syndrome (ARDS), a cause of substantial morbidity and mortality. Survival from ARDS is greater in children compared with adults. Whether developmental differences intrinsic to PEC barrier function contribute to
Maria Grazia Lampugnani et al.
Journal of cell science, 123(Pt 7), 1073-1080 (2010-03-25)
Little is known about the molecular mechanisms that regulate the organization of vascular lumen. In this paper we show that lumen formation correlates with endothelial polarization. Adherens junctions (AJs) and VE-cadherin (VEC, encoded by CDH5) are required for endothelial apicobasal

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service