The antibody detects endogenous levels of total KDM5A protein.
Immunogen
Synthetic peptide corresponding to a region derived from internal residues of human lysine (K)-specific demethylase 5A
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Molecular medicine reports, 16(3), 3573-3580 (2017-07-18)
The authors initially performed reverse transcription‑quantitative polymerase chain reaction to determine the expression profile of KDM5A in ovarian cancer tissues and adjacent normal tissue. Compared with adjacent normal tissue, it was identified that KDM5A was highly expressed in ovarian cancer
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.