Glucose-dependent insulinotropic polypeptide (GIP) or gastric inhibitory polypeptide is a 42-amino acid gastrointestinal regulatory peptide, expressed in K cells of the small intestine and in cells of the submandibular salivary gland. The gene encoding GIP is localized on human chromosome 17.
Specificity
The antibody detects endogenous levels of total GIP protein.
Immunogen
Fusion protein corresponding to a region derived from internal residues of human gastric inhibitory polypeptide
Biochem/physiol Actions
In the presence of glucose, glucose-dependent insulinotropic polypeptide (GIP) induces insulin secretion. It acts as an antagonist of the human GIP receptor. Aberrations in GIP are associated with visceral fat accumulation. The protein has been studied to have a protective function in obesity and atherosclerosis.
Features and Benefits
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Physical form
Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
The Gluco- and Liporegulatory and Vasodilatory Effects of Glucose-Dependent Insulinotropic Polypeptide (GIP) Are Abolished by an Antagonist of the Human GIP Receptor.
Asmar M, et al.
Diabetes (2017)
Rs46522 in the Ubiquitin-Conjugating Enzyme E2Z Gene Is Associated with the Risk of Coronary Artery Disease in Individuals of Chinese Han Population with Type 2 Diabetes.
Lu D, et al.
Journal of Diabetes Research (2017)
Common variants of GIP are associated with visceral fat accumulation in Japanese adults.
Nakayama K, et al.
American Journal of Physiology: Gastrointestinal and Liver Physiology, 307(11), G1108-G1114 (2014)
Incretin hormones receptor signaling plays the key role in antidiabetic potential of PTY-2 against STZ-induced pancreatitis.
Srivastava S, et al.
Biomedicine and Pharmacotherapy, 97, 330-338 (2018)
Cell-specific expression of the glucose-dependent insulinotropic polypeptide gene in a mouse neuroendocrine tumor cell line.
Boylan MO, et al.
The Journal of Biological Chemistry, 272(28), 17438-17443 (1997)
Questions
Reviews
★★★★★ No rating value
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.