Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4300865

Sigma-Aldrich

Anti-NEMF antibody produced in rabbit

affinity isolated antibody

Synonym(s):

NY-CO-1, SDCCAG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

123 kDa

species reactivity

human

concentration

1.8 mg/mL

technique(s)

immunohistochemistry: 1:25- 1:100
western blot: 1:200-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001288661.1

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NEMF(9147)

General description

Nuclear export mediator factor (NEMF), also known as Sdccag1 (Serologically Defined Colon Cancer Antigen 1) encodes a protein exhibiting tumor suppression property.It consists of four prominent domains: the N- and C-lobe connected by a coiled-coil to the middle (M) domain.

Specificity

The antibody detects endogenous levels of total NEMF protein.

Immunogen

Synthetic peptide corresponding to a region derived from internal residues of human nuclear export mediator factor

Application

Anti-NEMF antibody produced in rabbit is suitable for immunohistochemistry at a dilution of 1:25 - 1:100 and western blot at a dilution of 1:200-1:1000.

Biochem/physiol Actions

Nuclear export mediator factor (NEMF) is involved in the recruitment of Listerin to stalled ribosomes. It forms a complex with the nascent chain-containing 60S-Listerin and recruits and stabilizes Listerin′s N-terminal domain. It plays a vital role in mediating nuclear export. Inactivation of Sdccag1 expression leads to the transformation of human lung cancer cells.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Physical form

Rabbit IgG in pH7.3 PBS, 0.05% NaN3, 50% Glycerol.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sichen Shao et al.
Molecular cell, 57(3), 433-444 (2015-01-13)
During ribosome-associated quality control, stalled ribosomes are split into subunits and the 60S-housed nascent polypeptides are poly-ubiquitinated by Listerin. How this low-abundance ubiquitin ligase targets rare stall-generated 60S among numerous empty 60S is unknown. Here, we show that Listerin specificity
Xiaolin Bi et al.
Oncogene, 24(56), 8229-8239 (2005-08-17)
We previously showed that the Drosophila DNA binding homeodomain of Prospero included a 28 amino-acid sequence (HDA) that functions as a nuclear export signal. We describe here the identification of a protein we named Caliban, which can directly interact with
Assignment of the serologically defined colon cancer antigen 1 gene (SDCCAG1) to human chromosome band 14q22 by in situ hybridization.
D Carbonnelle et al.
Cytogenetics and cell genetics, 86(3-4), 248-249 (1999-11-27)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service