Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4300347

Sigma-Aldrich

Anti-ESR1 (Ab-167) antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZp686N23123 antibody produced in rabbit, Anti-ER antibody produced in rabbit, Anti-ESR antibody produced in rabbit, Anti-ESRA antibody produced in rabbit, Anti-estrogen receptor 1 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

~66 kDa

species reactivity

human

concentration

1 mg/mL

technique(s)

immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:50-1:100
indirect immunofluorescence: 1:100-1:200
western blot: 1:500-1:1000

isotype

IgG

immunogen sequence

(L-A-S-T-N)

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ESR1(2099)

General description

The estrogen receptor 1 (ESR1) gene is mapped to human chromosome 6q25.1-q25.2. The encoded protein belongs to the nuclear hormone receptor family. ESR1 is characterized with a transcription activation function-1 domain (AF-1), a DNA-binding domain (DBD), a hinge domain and a ligand-binding domain (LBD/AF-2).

Immunogen

Peptide sequence around aa. 165-169 (L-A-S-T-N), according to the protein ESR1.

Biochem/physiol Actions

Estrogen receptor 1 (ESR1) is involved in immune and inflammatory response. Aberrations in ESR1 results in Mayer-Rokitansky-Küster-Hauser syndrome that is associated with the congenital absence of the uterus and the upper two-thirds of the vagina. Polymorphism in the gene increases the risk of susceptibility to fractures. Elevated expression of the gene has been observed in breast cancer patients.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Target description

Nuclear hormone receptor. The steroid hormones and their receptors are involved in the regulation of eukaryotic gene expression and affect cellular proliferation and differentiation in target tissues.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide and 50% glycerol

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Activating ESR1 mutations in hormone-resistant metastatic breast cancer.
Robinson D R, et al.
Nature Genetics, 45(12), 1446-1446 (2013)
Differential genetic effects of ESR1 gene polymorphisms on osteoporosis outcomes.
Loannidis J P, et al.
JAMA : The Journal of the American Medical Association, 292(17), 2105-2114 (2004)
An array CGH based genomic instability index (G2I) is predictive of clinical outcome in breast cancer and reveals a subset of tumors without lymph node involvement but with poor prognosis.
Bonnet F, et al.
BMC Medical Genomics, 5(1), 54-54 (2012)
Estrogen receptor alpha (ESR1) gene amplification is frequent in breast cancer.
Holst F, et al.
Nature Genetics, 39(5), 655-655 (2007)
Assignment1 of the horse progesterone receptor (PGR) and estrogen receptor (ESR1) genes to horse chromosomes 7 and 31, respectively, by in situ hybridization.
Lear T L, et al.
Cytogenetic and genome research, 82(1-2), 110-111 (1998)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service