Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200834

Sigma-Aldrich

Anti-Porphyromonas gingivalis LPS antibody, Mouse monoclonal

clone PG-60, purified from hybridoma cell culture

Synonym(s):

Porphyromonas gingivalis LPS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

PG-60, monoclonal

description

Research area: Microbiome

form

liquid

species reactivity

Porphyromonas gingivalis

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 0.12-0.25 μg/mL using dead P.gingivalis bacteria.
indirect ELISA: 0.12-0.25 μg/mL using P. gingivalis LPS for coating.

isotype

IgG2b

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Porphyromonas gingivalis is a nonmotile, Gram-negative, rod-shaped, anaerobic pathogenic bacterium, belongs to the Porphyromonadaceae family, phylum Bacteroidetes class.1 P. gingivalis is involved in several pathogenic mechanisms including tissue colonization and destruction, as well as host defense activation.1-5 This bacterium is known for its ability to colonize at the oral cavity and is associated with public health concern. It can also be found in lung, liver, or splenic abscesses and the upper gastrointestinal tract and is related to atherosclerotic plaque disease, Cardiovascular diseases, Adverse pregnancy outcomes, Rheumatoid arthritis (RA), Meningitis or brain abscesses and Alzheimer′s disease.2-8

Specificity

Monoclonal Anti-Porphyromonas gingivalis LPS specifically recognizes P. gingivalis whole extract and P. gingivalis Lipopolysaccharides (LPS), the antibody has no cross reactivity with whole extract of Proteus mirabilis, Proteus vulgaris, E. coli, Pseudomonas aeruginosa, Shigella flexneri, Staphylococcus aureus or Salmonella enterica.

Immunogen

Porphyromonas gingivalis LPS

Application

The antibody is recommended to be used in various immunological techniques, including Immunoblot and ELISA. Detection of the P. gingivalis LPS bands by Immunoblotting is specifically inhibited by the immunogen.

Biochem/physiol Actions

P. gingivalis ability to aggregate in biofilms is considered a significant health risk by elevated resistance to host defense mechanisms and decreased susceptibility to conventional antimicrobials agents.9 P. gingivalis have several potential secreted and membranal virulence factors such as Gingipains proteolytic complex (Rgp and Kgp) which degrade various host proteins, Fimbriae (FimA and Mfa1) responsible for attachment of bacterial cells to host cell surfaces and Haemagglutinins (HagA) involved in adhesion and invasion of host cells.1-10 The host response to P. gingivalis infection may involve immunoglobulin degradation, inactivation of cytokines and their receptors, platelet aggregation, attenuation of neutrophil antibacterial activities and increasing vascular permeability, as well as, prevention of blood clotting.2  Extensive study from recent years, examine the influence of P. gingivalis LPS and P. gingivalis infections on RA disease. The production of citrullinated peptides by P. gingivalis virulence factors arginine gingipains (RgpA and RgpB) followed by peptidyl arginine deiminase enzyme (P.PAD) preferentially citrullinates C-terminal arginine result in host autoimmune response and generation of anti-citrullinated antibodies (ACPAs). Furthermore, in animal models P. gingivalis infection triggered an experimental autoimmune arthritis. 8,11-12

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Fata Moradali et al.
NPJ biofilms and microbiomes, 8(1), 53-53 (2022-07-07)
Microbial pathogens employ signaling systems through cyclic (di-) nucleotide monophosphates serving as second messengers to increase fitness during pathogenesis. However, signaling schemes via second messengers in Porphyromonas gingivalis, a key Gram-negative anaerobic oral pathogen, remain unknown. Here, we report that
Shirin Ghods et al.
Frontiers in cellular and infection microbiology, 14, 1418651-1418651 (2024-06-27)
This study unveils the intricate functional association between cyclic di-3',5'-adenylic acid (c-di-AMP) signaling, cellular bioenergetics, and the regulation of lipopolysaccharide (LPS) profile in Porphyromonas gingivalis, a Gram-negative obligate anaerobe considered as a keystone pathogen involved in the pathogenesis of chronic

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service