Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200833

Sigma-Aldrich

Anti-F. tularensis LPS antibody, Human-rabbit Chimeric monoclonal

clone TL1, purified recombinant antibody

Synonym(s):

Francisella tularensis LPS

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

antibody form

purified antibody

antibody product type

primary antibodies

clone

TL1, monoclonal

description

Research area: Microbiome

form

liquid

species reactivity

Francisella tularensis

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

ELISA: 0.5-1 μg/mL using Francisella tularensis LVS (live vaccine strain) extract.
immunoblotting: 1 μg/mL using Francisella tularensis LVS (live vaccine strain) extract.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Francisella tularensis is a Gram-negative bacteria that causes tularemia. Tularemia is a highly contagious and fatal zoonotic disease isolated from hundreds of animal species with severe clinical symptoms, including skin and gastrointestinal lesions and pneumonia.1-3 F. tularensis is listed as a category A biothreat agent by the CDC due to its aerosol and water-borne dissemination, high infectivity and it potential to high morbidity and mortality in humans. In nature F. tularensis can persist for long periods at low temperatures in animal carcasses, soil or water. There are three major F. tularensis group classified according to their geographic and virulence status.2-3

Specificity

Monoclonal Anti-F. tularensis LPS specifically recognizes Francisella tularensis and does not cross react with Escherichia coli bacteria.

Application

The antibody may be used in various immunochemical techniques including Immunoblot, Immunofluorescence, and ELISA. Detection of the F. tularensis LVS (live vaccine strain) in Immunoblotting is specifically inhibited by the immunogen. The recombinant antibody is composed of Human Fc chain and variable regions from rabbit origin, therefor it is recommended to use Anti-Human Fc antibody as the secondary antibody for detection.

Biochem/physiol Actions

F. tularensis virulence is unique and researchers are trying to understand the bacteria pathogenesis, mechanism of action and discover its virulence factors since it does not appear to secrete any toxins.2-4 Furthermore, its′ LPS has an unusual structure and is ~1000-fold less potent than most Gram-negative bacteria LPS and does not activate cells via Toll-like receptor 4 (TLR4). 3-4 Hence F. tularensis LPS can be use as good target for generation of specific antibodies and potential vaccine for F. tularensis infections.1,5

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Adva Mechaly et al.
Scientific reports, 9(1), 11418-11418 (2019-08-08)
Francisella tularensis (Ft), the causative agent of lethal tularemia, is classified as a category A biological warfare threat agent. While Ft infection is treatable by antibiotics, many failed antibiotic treatments were reported, highlighting the need for effective new treatments. It

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service