Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200831

Sigma-Aldrich

Anti- Staphylococcal Enterotoxin B-Peroxidase antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Enterotoxin type B, SEB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

horse radish peroxidase (HRP) conjugate

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

description

Research area: Microbiome

form

lyophilized powder

mol wt

25-30  kDa

species reactivity

Staphylococcus aureus

concentration

~4 mg/mL

technique(s)

direct ELISA: 1:8,000-1:16,000 using 1 μg/mL Staphylococcal Enterotoxin B for coating.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Staphylococcus aureus is a Gram positive bacterium, that cause diseases in human population including food poisoning and toxic shock syndrome.1 S. aureus express several secreted virulence factors, such as various enzymes, cytotoxins, exotoxins, and exfoliative toxins.1 Exotoxins include more than 20 serologically classified staphylococcal enterotoxins (SEs), the best characterize are the SEs A, B through V and toxic shock syndrome toxin-1 (TSST-1), these enterotoxins are similar in activity, sequence, structure and molecular weight (~25-30 kDa).1

Specificity

Anti- Staphylococcal Enterotoxin B- peroxidase antibody specifically recognizes Staphylococcal Enterotoxin B (SEB) and has no cross reactivity with Staphylococcal Enterotoxin A (SEA), Cholera Toxin or Pseudomonas Exotoxin A.

Immunogen

purified Enterotoxin B from Staphylococcus aureus

Application

The antibody may be used in various immunochemical techniques including ELISA.

Biochem/physiol Actions

SEA and SEB are known as superantigens due to their ability to bind class II MHC molecules on antigen presenting cells and stimulate extensive T-cells activation followed by massive cytokine release leading to an acute toxic shock.1-4

SE proteins have a significant resistance to heat and acid, it is also resistant to gastrointestinal proteases including pepsin, trypsin, rennin and papain. Thus, killing the S. aureus bacteria may not be sufficient to eliminate these superantigens risk from causing food poisoning.

Physical form

Supplied as a lyophilized powder.

Storage and Stability

Store the lyophilized product at 2–8 °C. For extended storage after reconstitution, keep at –20 °C in working aliquots. Avoid repeated freeze-thaw cycles. For continuous use after reconstitution, keep at 2–8 °C for up to 1 month. Solutions at working dilution should be discarded if not used within 12 hours.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.
Unless otherwise stated in our catalog  our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

11 - Combustible Solids

wgk_germany

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service