Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200825

Sigma-Aldrich

Anti-O-spanin antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Outer membrane lipoprotein Rz1 from lambdoid prophage DLP12, Prophage outer membrane lipoprotein RzoD

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

description

Research area: Microbiome

form

buffered aqueous solution

mol wt

~20 kDa

species reactivity

E. coli

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoblotting: 1:500-1:1000 using E. coli lysate.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Escherichia coli K12 ... rzoD(1450242)

General description

E. coli o-spanin, also known as Outer membrane lipoprotein Rz1 from lambdoid prophage DLP12, RzoD or RzoR, is originated from the defective Coliphage lambda DLP12 genome carried by E. coli and present in nearly all lambdoid phages and many additional bacteriophages. DLP12 cassette in E. coli contains four lysis proteins ORFs; a holin (S) through which the endolysin (R) and spanin (Rz/Rz1) escape into the periplasmic space. Two Spanin cell-lysis proteins, o-spanin an outer membrane lipoprotein and i-spanin (rzpD) an integral inner membrane protein are homologues of the Lambda phage RZ1 and RZ genes respectively, the Rz1 gene is uniquely located within the +1 reading frame of Rz.

Specificity

Anti-o-spanin antibody specifically recognizes the Outer membrane lipoprotein subunit o-spanin from E. coli and Shigella flexneri and does not cross react with Proteus mirabilis

Immunogen

Peptide: E.coli O-spanin. CAWIMQPPPDWQTPLNG

Application

The antibody may be used in various immunochemical techniques including Immunoblotting (predicted ~6.5kDa observed ~20 kDa due to oligomerization). Detection of the o-spanin band by Immunoblotting is specifically inhibited by the immunizing peptide.

Biochem/physiol Actions

Spanins are conserved among lambdoid phages and comprise the final step of lysis in lambda phage infections of E. coli. These proteins form heterotetramer complex composed of two homodimers linked by homotypic intermolecular disulfide bonds, span the periplasm and bridge the cell envelope thus causing fusion of the inner and outer membranes and disruption of the bacterium membrane.The DLP12 lysis genes play an important, yet to be identified role in the bacterial cell wall maintenance and biofilm formation. This was shown using mutated strains lacking essD, ybcS and rzpD/rzoD that were unable to form biofilms and had significant lower aggregation as compared with wild-type. These finding suggests that defective prophage genes are affecting host cell physiology contribute to host fitness and survival, biofilm dispersion, enhanced biofilm traits such as antibiotic resistance, attachment, phage tolerance and metabolite recycling.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For continuous use, store at 2-8°C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12
hours.

Disclaimer

Unless otherwise stated in our catalog our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service