Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200789

Sigma-Aldrich

Anti-Human Lambda Light Chains (Bound and Free)-Peroxidase antibody, Mouse monoclonal

clone HP-6054, purified from hybridoma cell culture

Synonym(s):

Anti-Human immunoglobulin lambda light chain

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.46

biological source

mouse

conjugate

peroxidase conjugate

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

HP-6054, monoclonal

form

lyophilized powder

species reactivity

human

concentration

~2 mg/mL

technique(s)

direct ELISA: 1:16,000-1:32,000 using using 2 μg/ml human IgG lambda light chains for coating.

isotype

IgG2a

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Anti-Human Lambda Light Chains (Bound and Free)- Peroxidase antibody, Mouse monoclonal (mouse IgG2a isotype) is derived from the HP-6054 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from a mouse immunized with purified human IgG myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads. Immunoglobulins are composed of two heavy and two light polypeptide chains held together by noncovalent forces and usually by interchain disulfide bridges. The various types of human (and other mammalian) immunoglobulins contain one of the two existing light chain types, either κ or λ in which multiple structural differences are reflected in antigenic variety, mainly the N-terminal (variable) domain of the chains.

Immunogen

Purified human IgG myeloma proteins covalently coupled to polyaminostyrene (PAS) microbeads.

Application

Direct ELISA: a working dilution of 1:16,000-1:32,000 is recommended using 2 μg/mL human IgG lambda light chains for coating.

Biochem/physiol Actions

In monoclonal disorders, such as myeloma and macroglobulinemia, an increase in the level of a single immunoglobulin class can be accompanied by disproportionate increase in either λ or κ light chains. In many cases of B cell malignancy there is an increased production of light chains which are not accompanied with heavy chains. These circulate in the various body fluids (blood, cerebral spinal fluid, and tissues) and are also found in enormous amounts as free molecules in the urine (Bence Jones paraproteins).

Physical form

Supplied as a lyophilized powder.

Other Notes

In order to obtain best results in different techniques and preparations we recommend determining optimal working concentration by titration test.

Disclaimer

For U.S. Customers: Contains mercury; Do not place in trash - dispose according to local, state, or federal laws.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

13 - Non Combustible Solids

wgk_germany

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service