Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200771

Sigma-Aldrich

Anti-Surfactant Protein D antibody, Mouse monoclonal

clone 10F6E12, purified from hybridoma cell culture

Synonym(s):

Anti-Collectin-7, Anti-Lung surfactant protein D, Anti-PSP-D, Anti-Pulmonary surfactant-associated protein D, Anti-SP-D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

10F6E12, monoclonal

form

buffered aqueous solution

mol wt

~43 kDa

species reactivity

mouse, human

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using mouse lung extract

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SFTPD(6441)
mouse ... Sftpd(20390)

General description

Surfactant Protein D (SFTPD) belongs to the collectin family of multimeric glycoproteins and exists as oligomeric structure or a trimeric subunit. The human SFTPD corresponds to a molecular weight of 150kDa and the trimer is close to 620 kDa. Anti-Surfactant Protein D antibody, Mouse monoclonal, (mouse IgG1 isotype) is derived from the 10F6E12 hybridoma, produced by the fusion of mouse myeloma cells and splenocytes from mouse immunized with mouse recombinant SP-D.
Surfactant Protein D (SP-D) contains an N-terminal cysteine-rich domain, a collagenous domain, a neck region and a C-terminal carbohydrate recognition domain (CRD). The gene is located on human chromosome 14. SP-D is expressed in lungs and kidney.

Specificity

Anti-Surfactant Protein D antibody, Mouse monoclonal recognizes SP-D from human and mouse origin.

Immunogen

Mouse recombinant SP-D

Application

Anti-Surfactant Protein D antibody, Mouse monoclonal may be used in immunoblotting.

Biochem/physiol Actions

Surfactant Protein D (SFTPD) imparts pulmonary defense during fungal infection. It favors pulmonary inflammation. It recognizes the surface lipopolysaccharide (LPS) of pathogens and acts as first-line host defense molecule. It also recognizes the viral glycoprotein hemagglutinin (HA) present in viral surface. Polymorphism in the SFTPD gene may lead to decreased levels of the protein and high risk for chronic obstructive pulmonary disease (COPD).
Surfactant Protein D (SP-D) plays a critical role in the innate host defense and the modulation of inflammatory responses in infectious diseases. It aids the uptake and clearance of pathogens and apoptotic cells. SP-D maintains surfactant homeostasis. It reduces the surface tension at the air-liquid interface and blocks the lung from collapsing.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular mechanisms of inhibition of influenza by surfactant protein D revealed by large-scale molecular dynamics simulation
Goh BC, et al.
Biochemistry, 52(47), 8527-8538 (2013)
Single nucleotide polymorphisms in collagenous lectins and other innate immune genes in pigs with common infectious diseases
Keirstead ND, et al.
Veterinary Immunology and Immunopathology, 142(1-2), 1-13 (2011)
Genetic loci associated with chronic obstructive pulmonary disease overlap with loci for lung function and pulmonary fibrosis
Hobbs BD, et al.
Nature Genetics, 49(3), 426-426 (2017)
Antifungal activities of surfactant protein D in an environment closely mimicking the lung lining
Ordonez SR, et al.
Molecular Immunology, 105, 260-269 (2019)
Lung remodeling in aging surfactant protein D deficient mice
Schneider JP, et al.
Annals of Anatomy = Anatomischer Anzeiger : Official Organ of the Anatomische Gesellschaft, 211, 158-175 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service