Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200765

Sigma-Aldrich

Anti-tau antibody, Mouse monoclonal

clone tu53, purified from hybridoma cell culture

Synonym(s):

Anti-Microtubule-associated protein tau (MART), Anti-Neurofibrillary tangle protein, Anti-Paired helical filament-tau (PHF-tau), Anti-Tau-F

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.41

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

tu53, monoclonal

form

buffered aqueous solution

mol wt

46-60 kDa

species reactivity

human, mouse

concentration

~1 mg/mL

technique(s)

immunoblotting: 5-10 μg/mL using mouse brain tissue extract
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MAPT(4137)

General description

Anti-tau antibody, mouse monoclonal (mouse IgG1 isotype) is derived from the tu53 hybridoma produced by the fusion of mouse myeloma cells and splenocytes from BALB /C mice. Tau, also known as microtubule-associated protein tau (MAPT), belongs to microtubule-associated proteins (MAP) family. It consists of six isoforms in the neurons of the adult human brain. MAPT gene is located on the human chromosome at 17q21.31.

Specificity

Anti-tau antibody, Mouse monoclonal specifically recognizes human and mouse tau protein. The antibody recognizes all six isoforms of human tau.

Immunogen

Recombinant human tau protein expressed in HEK-293 cells

Application

Anti-tau antibody, mouse monoclonal may be used in immunoblotting and immunoprecipitation.

Biochem/physiol Actions

Tau loses its microtubule-binding activity and aggregates into paired helical filaments (PHFs) during neurodegenerative disorders. It can undergo modifications such as hyperphosphorylation, nonenzymatic glycosylation and acetylation, which result in the generation of aberrant aggregates such as found in neurofibrillary tangles (NFTs) in Alzheimer′s disease. It is present abundantly in intraneuronal and glial fibrillar lesions formed during neurodegenerative disorders such as tauopathies and Alzheimer′s disease.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

17q21. 31 duplication causes prominent tau-related dementia with increased MAPT expression
Le Guennec K, et al.
Molecular Psychiatry, 22(8), 1119-1125 (2017)
Tau protein isoforms, phosphorylation and role in neurodegenerative disorders
Buee L, et al.
Brain Research Reviews, 33(1), 95-130 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service