SAB4200754
Anti-IL6 antibody produced in rabbit
IgG fraction of antiserum, buffered aqueous solution
Synonym(s):
Anti-B-cell stimulatory factor 2 (BSF-2), Anti-CTL differentiation factor (CDF), Anti-Hybridoma growth factor, Anti-Interferon beta-2 (IFN-beta-2), Anti-Interleukin-6
About This Item
biological source
rabbit
antibody form
IgG fraction of antiserum
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
~23 kDa
species reactivity
human
concentration
~1.0 mg/mL
technique(s)
immunoblotting: 1:2,000-1:4,000 using human osteosarcoma MG-63 concentrated supernatant treated with IL1ß
immunoprecipitation (IP): 1-2.5 μL using human osteosarcoma MG-63 concentrated supernatant treated with IL1ß
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... IL6(3569)
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service