SAB4200727
Anti-AKT2 antibody, Mouse monoclonal
clone AKT2(104), purified from hybridoma cell culture
Synonym(s):
Anti-PKB beta, Anti-Protein kinase Akt-2, Anti-RAC-beta serine/threonine-protein kinase
About This Item
biological source
mouse
antibody form
purified from hybridoma cell culture
antibody product type
primary antibodies
clone
AKT2(104), monoclonal
form
buffered aqueous solution
mol wt
~56 kDa
species reactivity
rat, mouse, human
concentration
~1.0 mg/mL
technique(s)
immunoblotting: 10-20 μg/mL using human T lymphocyte Jurkat cell extract
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... AKT2(208)
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service