Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200687

Sigma-Aldrich

Monoclonal Anti-NCOR2/SMRT antibody produced in mouse

clone SMRT 5.1, purified from hybridoma cell culture

Synonym(s):

CTG26, N-CoR2, SMAP270, SMRT, SMRTE, SMRTE-tau, TNRC14, TRAC, TRAC-1, TRAC1, nuclear receptor corepressor 2

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$542.02

$542.02

List Price$657.00Save 18%
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

Select a Size

Change View
100 μL
$542.02

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$542.02

List Price$657.00Save 18%
Web-Only Promotion

Available to ship onApril 29, 2025Details


Request a Bulk Order

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

SMRT 5.1, monoclonal

form

buffered aqueous solution

mol wt

antigen ~270 kDa

species reactivity

human, mouse, canine, bovine, rat

concentration

~1 mg/mL

technique(s)

immunoblotting: 2-4 μg/mL using nuclear extract of human HeLa cells.
immunofluorescence: 5-10 μg/mL using human HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CHD5(26038)

General description

Monoclonal Anti-NCOR2/SMRT (mouse IgG1 isotype) is derived from the hybridoma SMRT5.1 produced by the fusion of mouse myeloma cells and splenocytes from mice immunized with the C-terminal region of GST-tagged SMRT protein. Silencing mediator for retinoid or thyroid-hormone receptor or nuclear receptor co-repressor 2 (NCOR2) is a large protein with multiple functional domains. It comprises the repression domain, Swi3, Ada2, NCoR1, and TFIIB (SANT)-like domain and a receptor interaction domain (RID). NCOR2 gene is mapped to human chromosome 12q24.31.

Specificity

Monoclonal Anti-NCOR2/SMRT recognizes human NCOR2/SMRT protein

Immunogen

the C-terminal region of GST-tagged SMRT protein

Application

Monoclonal Anti-NCOR2/SMRT antibody produced in mouse may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Silencing mediator for retinoid or thyroid-hormone receptor (SMRT) also known as SMAP270 or T3 receptor associating cofactor 1 (TRAC-1), serves as a transcription repressive co-regulatory factor for multiple pathways. A few to mention including inflammation, metabolism, circadian rhythms, developmental and homeostatic processes. Also, SMRT is essential for histone deacetylase 3 (HDAC3) activity that is required for chromatin structure and genome stability. SMRT protein is a part of NCOR/SMRT-mediated repression pathway together with the nuclear receptor corepressor 1 (NCOR1). They play pathophysiological roles in acute promyelocytic leukemia (APL), acute myelogenous leukemia (AML), multiple myeloma, Rett syndrome and deafness. NCoR2 is a target for microRNAs associated with glioblastoma.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

nwg

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Y Huang et al.
Genes & development, 14(1), 45-54 (2000-01-20)
Transcriptional repression mediated by corepressors N-CoR and SMRT is a critical function of nuclear hormone receptors, and is dysregulated in human myeloid leukemias. At the present time, these corepressors are thought to act exclusively through an mSin3/HDAC1 complex. Surprisingly, however
Adrienne Mottis et al.
Genes & development, 27(8), 819-835 (2013-05-01)
Epigenetic regulation of gene expression is strongly influenced by the accessibility of nucleosomal DNA or the state of chromatin compaction. In this context, coregulators, including both coactivators and corepressors, are pivotal intermediates that bridge chromatin-modifying enzymes and transcription factors. NCoR1
Amanda Shea et al.
Cancer medicine, 5(8), 1917-1946 (2016-06-11)
Glioblastoma multiforme (GBM) is the most common and lethal cancer of the adult brain, remaining incurable with a median survival time of only 15 months. In an effort to identify new targets for GBM diagnostics and therapeutics, recent studies have focused
Genomic characterization of spleens in patients with myelofibrosis.
Eran Zimran et al.
Haematologica, 103(10), e446-e449 (2018-05-12)
Pushpankur Ghoshal et al.
Cancer research, 69(10), 4380-4387 (2009-05-07)
Multiple myeloma (MM) is a clonal B-cell neoplasm that accounts for 10% of all malignant hematologic neoplasms and that affects terminally differentiated B cells (i.e., plasma cells). It is now well recognized that the cytokine interleukin-6 (IL-6) is a major

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service