Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200675

Sigma-Aldrich

Monoclonal Anti-Multimerin-2 antibody produced in mouse

clone MM-5, purified from hybridoma cell culture

Synonym(s):

EMILIN-3, EMILIN3, ENDOGLYX-1, MMRN2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

MM-5, monoclonal

form

buffered aqueous solution

mol wt

antigen ~104 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 5-10 μg/mL using whole extract of human HepG2 cells.
immunofluorescence: 2-5 μg/mL using human HeLa cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MMRN2(79812)

General description

Monoclonal Anti-Multimerin-2 (mouse IgG1 isotype) is derived from the hybridoma MM-5 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Multimerin-2 (MMRN2) belongs to the EMI domain endowed (EDEN) protein family. It is located in the blood vessels and is a glycoprotein. MMRN2 gene is mapped on the human chromosome at 10q23.2.
Multimerin-2 (MMRN2) is an extracellular matrix glycoprotein. The protein contains a signal sequence, an amino-terminal EMI (cysteine-rich) domain and a carboxyl-terminal C1q (complement 1)-like domain. MMRN2 is also referred to as endoglyx-1.

Specificity

Monoclonal Anti-Multimerin-2 recognizes human Multimerin-2.

Immunogen

a recombinant protein of human Multimerin-2

Application

Monoclonal Anti-Multimerin-2 antibody produced in mouse may be used in immunoblotting and immunofluorescence.

Biochem/physiol Actions

Multimerin-2 (MMRN2) negatively regulates the angiogenesis regulators vascular endothelial growth factor receptor 1 & 2 (VEGFR 1 & 2). It plays a role in the maintenance of blood vessels homeostasis, regulation of endothelial cell (EC) function and hence tumor growth arrest.
Multimerin-2 (MMRN2) regulates endothelial cell migration and neo-angiogenesis through interaction with VEGF-A (vascular endothelial growth factor-A).

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C1q and its growing family
Ghai R, et al.
Immunobiology, 212(4-5), 253-266 (2007)
Multimerin-2 maintains vascular stability and permeability
Pellicani R, et al.
Matrix Biology, 87(12), 11-25 (2020)
Loss of Multimerin-2 and EMILIN-2 expression in gastric cancer associate with altered angiogenesis
Andreuzzi E, et al.
International Journal of Molecular Sciences, 19(12), 3983-3983 (2018)
S Christian et al.
The Journal of biological chemistry, 276(51), 48588-48595 (2001-09-18)
EndoGlyx-1, the antigen identified with the monoclonal antibody H572, is a pan-endothelial human cell surface glycoprotein complex composed of four different disulfide-bonded protein species with an apparent molecular mass of approximately 500 kDa. Here, we report the purification and peptide
E Lorenzon et al.
Oncogene, 31(26), 3136-3147 (2011-10-25)
MULTIMERIN2 (MMRN2), also known as Endoglyx-1, is an extracellular matrix glycoprotein whose function has so far remained elusive. Given its specific localization in tight association with the endothelium we hypothesized that this protein could modulate neo-angiogenesis. By multiple assays we

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service