Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

SAB4200674

Sigma-Aldrich

Monoclonal Anti-PPIAL4 antibody produced in mouse

clone PP-8, purified from hybridoma cell culture

Synonym(s):

COAS-2, COAS2, PPIAL4, PPIAL4A, PPIAL4B, peptidylprolyl isomerase A (cyclophilin A)-like 4B

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

PP-8, monoclonal

form

buffered aqueous solution

mol wt

antigen ~18 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 2.5-5 μg/mL using whole extract of human HepG2 cells.

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Monoclonal Anti-PPIAL4 (mouse IgG1 isotype) is derived from the hybridoma PP-8 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice. Peptidyl-Prolyl cis-trans isomerase A-like 4A/B/C (PPIAL4B) is a novel member of the cyclosporin-binding peptidyl-prolyl isomerase family.
The gene PPIAL4A (peptidylprolyl isomerase A like 4A) is mapped to human chromosome 1q21.

Specificity

Monoclonal Anti-PPIAL4 recognizes human PPIAL4B.

Immunogen

a recombinant protein of human PPIAL4B

Application

Monoclonal Anti-PPIAL4 antibody produced in mouse may be used in immunoblotting.

Biochem/physiol Actions

PPIAL4A (peptidylprolyl isomerase A like 4A) is a propyl isomerase which is required for folding of the proteins. In bone and soft tissue tumor, PPIAL4A exhibits high copy number.
Peptidyl-Prolyl cis-trans isomerase A-like 4A/B/C (PPIAL4B) is over-expressed almost exclusively in aggressive metastatic or chemotherapy-resistant tumors. Elevated levels of PPIAL4B are observed in bladder cancer, hepatocellular carcinomas, breast carcinomas and sarcomas.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2–8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Malin Nilsson et al.
International journal of cancer, 109(3), 363-369 (2004-02-13)
Amplifications and gains involving 1q are common abnormalities in solid tumors. Recently, an amplicon originating from 1q21-23, containing the candidate oncogenes COAS1, COAS2 and COAS3 (Chromosome One Amplified Sequence) was identified. The presence, distribution and copy number level of extra
Kosuke Yoshihara et al.
Genes, chromosomes & cancer, 50(3), 167-177 (2011-01-08)
We investigated characteristics of germline copy number variations (CNV) in BRCA1-associated ovarian cancer patients by comparing them to CNVs present in sporadic ovarian cancer patients. Germline CNVs in 51 BRCA1-associated, 33 sporadic ovarian cancer patients, and 47 healthy women were
Parveen Bhatti et al.
Chronobiology international, 32(1), 103-112 (2014-09-05)
The negative health effects of shift work, including carcinogenesis, may be mediated by changes in DNA methylation, particularly in the circadian genes. Using the Infinium HumanMethylation450 Bead Array (Illumina, San Diego, CA), we compared genome-wide methylation between 65 actively working

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service