Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200659

Sigma-Aldrich

Monoclonal Anti-Glucokinase (GCK) antibody produced in mouse

clone GCK-20, purified from hybridoma cell culture

Synonym(s):

FGQTL3, GK, GLK, HHF3, HK4, HKIV, HXKP, LGLK, MODY2, glucokinase (hexokinase 4)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GCK-20, monoclonal

form

buffered aqueous solution

mol wt

~50 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 10-20 μg/test using A549 cells
immunoblotting: 1.25-2.5 μg/mL using whole extract of HepG2 cells.
immunohistochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GCK(2645)

General description

The gene encoding glucokinase (GCK) is alternatively spliced to generate three different forms of the enzyme; one found in the pancreas and two found in the liver. The enzyme is expressed as a monomer and possesses one glucose-binding site. The gene encoding GCK is localized on human chromosome 7.[1] Recombinant human liver glucokinase (Accession# NP_277042)) has a C-terminal FLAG tag and has 474 amino acid residues.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal region of human Glucokinase

Biochem/physiol Actions

Glucose is phosphorylated to glucose-6-phosphate by glucokinases. The main function of glucokinase (GCK) is to regulate carbohydrate metabolism. The enzyme triggers the release of insulin in β-cells of pancreas. It can be useful for studies including enzyme kinetics, activator screening and kinase selectivity. Mutation in the gene encoding this protein is associated with a rare form of diabetes and the protein may also have a role in type 2 diabetes.[1]

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dual allosteric activation mechanisms in monomeric human glucokinase.
Whittington AC
Proceedings of the National Academy of Sciences of the USA, 112(37), 11553-11558 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service