Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200655

Sigma-Aldrich

Monoclonal Anti-NCAPH2 antibody produced in rat

clone 5F2G4, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
51111800
NACRES:
NA.41

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5F2G4, monoclonal

form

buffered aqueous solution

species reactivity

human, rat, mouse

concentration

~1 mg/mL

technique(s)

flow cytometry: 1-2 μg/test using using HeLa cells
immunoblotting: 2-4 μg/mL using using whole extracts of HeLa cells
immunofluorescence: 0.25-0.5 μg/mL using using HeLa cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Non-SMC condensin II complex subunit H2 (NCAPH2) is one of the subunits of the condensin II complex. The gene encoding it is localized on human chromosome 22q13.

Immunogen

a human NCAPH2 (full-length) recombinant fusion protein

Application

Monoclonal Anti-NCAPH2 antibody produced in rat has been used in immunostaining.

Biochem/physiol Actions

Non-SMC condensin II complex subunit H2 (NCAPH2) binds to mitotic chromosomes and is associated with mitosis. Mutations in the gene encoding NCAPH2 have been shown to impair T-cell functions in mice. The NCAPH2 gene is associated with mitosis and meiosis, which predicts that its methylation influences apoptosis of neural cells. NCAPH2 methylation causes loss of hippocampal function and cognitive dysfunction leading to progressive dementia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Condensin II and GAIT complexes cooperate to restrict LINE-1 retrotransposition in epithelial cells.
Jacqueline R W, et al.
PLoS ONE, 13(10), e1007051-e1007051 (2017)
Dynamic Organization of Chromatin Domains Revealed by Super-Resolution Live-Cell Imaging
Tadasu N, et al.
Molecular Cell, 67(2), 282-293 (2017)
Chih-Ping Chen et al.
European journal of medical genetics, 53(5), 329-332 (2010-06-15)
We report a 5-year-old boy with mental retardation, autistic features, epilepsy, developmental delay, atopic dermatitis and abnormal immunological findings, carrying a 7.9 Mb de novo deletion of chromosome 22q13.2→qter. This region contains the SHANK3, NCAPH2 and CYP2D6 genes which are
A novel role for the condensin II complex in cellular senescence.
Yokoyama Y
Cell Cycle, 14(13), 2160-2170 (2015)
Angelo Theodoratos et al.
The FEBS journal, 279(8), 1422-1432 (2012-02-16)
Condensins I and II are five-protein complexes that are important for the condensation of chromatin. They are essential for mitosis and important for regulating gene expression during interphase. Here, we investigated the transcription and translation of the mouse Ncaph2 gene

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service