Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200653

Sigma-Aldrich

Monoclonal Anti-GLUT4/SLC2A4 antibody produced in mouse

clone Glut2.6, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

Glut2.6, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

immunoblotting: 4-8 μg/mL using using whole extracts of HeLa cells.
immunofluorescence: 5-10 μg/mL using using HeLa cells

isotype

IgM

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GLUT4(6517)

General description

Solute carrier family 2 member 4 (SLC2A4) is also known as glucose transporter-4 (GLUT4). It is highly expressed in adipose tissue and skeletal muscles. The gene encoding SLC2A4 is localized on human chromosome 17p13.

Immunogen

a synthetic peptide corresponding to a sequence at the C-terminal region of human GLUT4, conjugated to KLH.

Biochem/physiol Actions

Solute carrier family 2 member 4 (SLC2A4) transports glucose into cells in response to insulin. Levels of this protein have been shown to be reduced in type 2 diabetes.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G I Bell et al.
Diabetes, 38(8), 1072-1075 (1989-08-01)
Glucose uptake by heart, skeletal muscle, and adipose tissue is acutely regulated by insulin, which stimulates facilitative glucose transport, at least in part, by promoting the translocation of transporters from an intracellular pool to the plasma membrane. cDNAs encoding the
Malgorzata Malodobra-Mazur et al.
Gene, 576(1 Pt 3), 499-504 (2015-11-04)
The glucose uptake by numerous cells like adipocytes or skeletal muscle cells requires activation of specific proteins called glucose transporters. GLUT4 content and its activation as well as SLC2A4 gene expression level are decreased in patients with type 2 diabetes.
Wei-Feng Shan et al.
Journal of Zhejiang University. Science. B, 12(8), 677-682 (2011-07-29)
Decreased glucose tolerance and diabetes are frequently observed in advanced liver cirrhosis patients and may be related to insulin resistance. Glucose transporter-4 (GLUT4), one of the most important glucose transporters, plays a key role in the development of type 2

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service