Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200640

Sigma-Aldrich

Monoclonal Anti-CHD5 antibody produced in rat

clone 5A10, purified from hybridoma cell culture

Synonym(s):

Anti-ATP−dependent helicase CHD5, Anti-CHD−5, Anti-CHD5, Anti-RP1−233K16.2, Anti-chromodomain helicase DNA binding protein 5, Anti-chromodomain−helicase−DNA−binding protein 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rat

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

5A10, monoclonal

form

buffered aqueous solution

mol wt

~250 kDa

species reactivity

monkey, mouse, human

concentration

~1 mg/mL

technique(s)

flow cytometry: 2.5-5 μg/test using using HeLa cells.
immunoblotting: 1.0-2.0 μg/mL using using whole extracts of L929 cells.
immunofluorescence: 5-10 μg/mL using using HeLa cells.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

Monkey ... CHD5(720712)
human ... CHD5(26038)
mouse ... 269610(Chd5)

General description

Monoclonal Anti-CHD5 (rat IgG2a isotype) is derived from the hybridoma 5A10 produced by the fusion of mouse myeloma cells and lymph node cells from rat immunized with a synthetic peptide corresponding to a sequence at the C-terminal region of human CHD5. Chromodomain helicase DNA(CHD) binding protein 5 (CHD5) is an ATP-dependent chromatin-remodeling factor. It is mainly expressed in neuronal tissues. CHD5 is a member of CHD family of proteins that is characterized with histone binding chromo (chromatin organization modifier) domains and sucrose non-fermentable 2 (SNF2)-related helicase/ATPase domains.

Immunogen

synthetic peptide corresponding to a sequence at the C-terminal region of human CHD5 .

Application

Monoclonal Anti-CHD5 antibody produced in rat has been used in various immunochemical techniques including:
  • immunoblotting
  • flow cytometry
  • immunocytochemistry
  • immunohistochemistry

Biochem/physiol Actions

Chromodomain helicase DNA binding protein 5 (CHD5) regulates genes involved in neuronal function, cell-cycle control, and chromatin remodeling. Interestingly, CHD5 is frequently deleted in neuroblastoma and thus suggested as a tumor suppressor gene. Furthermore, non-conformist CHD5 promoter methylation was identified in several primary tumors.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mechanisms of CHD5 Inactivation in neuroblastomas
Koyama H, et al.
Clinical Cancer Research, 18(6), 15881-11597 (2012)
Kai-Oliver Henrich et al.
Cancer research, 72(23), 6079-6088 (2012-11-23)
A broad range of human malignancies is associated with nonrandom 1p36 deletions, suggesting the existence of tumor suppressors encoded in this region. Evidence for tumor-specific inactivation of 1p36 genes in the classic "two-hit" manner is scarce; however, many tumor suppressors
Jinhua Quan et al.
PloS one, 9(9), e108066-e108066 (2014-09-24)
Loss of the chromatin remodeling ATPase CHD5 has been linked to the progression of neuroblastoma tumors, yet the underlying mechanisms behind the tumor suppressor role of CHD5 are unknown. In this study, we purified the human CHD5 complex and found

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service