Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200636

Sigma-Aldrich

Monoclonal Anti-CDA antibody produced in mouse

clone CDA-40, purified from hybridoma cell culture

Synonym(s):

CDD, Cytidine aminohydrolase, cytidine deaminase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

CDA-40, monoclonal

form

buffered aqueous solution

mol wt

~16 kDa

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using using HeLa cells.
immunoblotting: 0.5-1 μg/mL using whole extracts of HeLa cells.
immunofluorescence: 4-8 μg/mL using using HeLa cells.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CDA(978)

General description

Monoclonal Anti-CDA (mouse IgG2a isotype) is derived from the hybridoma CDA-40 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide corresponding to an internal sequence of human CDA conjugated to KLH. CDA (Cytidine deaminase) also known as cytidine aminohydrolase or CDD. The enzyme is active only as an intact tetramer, since three different monomers concur to the formation of each of the four active sites.

Immunogen

synthetic peptide corresponding to an internal sequence of human CDA .

Application

Monoclonal Anti-CDA antibody may be used in several immunochemical techniques including:
  • immunoblotting (~16 kDa)
  • flow cytometry
  • immunocytochemistry

Biochem/physiol Actions

Cytidine deaminase (CDA) is an intracellular enzyme of the pyrimidine salvage pathways. It catalyzes that deamination of cytidine and deoxycytidine to uridine and deoxyuridine, respectively. In addition, It also catalyzes the deamination of cytosine nucleoside analogues such as ARA-C (cytosine arabinoside) and 5-AZA-CdR (5-aza-2′-deoxycytidine) used in chemotherapy, leading to the loss of their pharmacological activity. CDA inhibitors such as tetrahydrouridine (THU) are interesting therapeutically for use in combination with chemotherapy drugs in patients with leukemia, to enhance drugs activity.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Laliberté et al.
Cancer chemotherapy and pharmacology, 30(1), 7-11 (1992-01-01)
Deamination of the nucleoside analogues ARA-C and 5-AZA-CdR by CR deaminase results in a loss of antileukemic activity. To prevent the inactivation of these analogues, inhibitors of CR deaminase may prove to be useful agents. In the present study we
Nucleosidic DNA demethylating epigenetic drugs--a comprehensive review from discovery to clinic
Agrawal K, et al.
Pharmacology & Therapeutics, 188(1), 45-79 (1992)
Stefano Costanzi et al.
ChemMedChem, 6(8), 1452-1458 (2011-05-21)
Cytidine deaminase (EC 3.5.4.5, CDA), an enzyme of the pyrimidine salvage pathways, is responsible for the degradation and inactivation of several cytidine-based antitumor drugs such as cytarabine, gemcitabine, decitabine, and azacytidine. Thus, CDA inhibitors are highly sought after as compounds to

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service