SAB4200635
Monoclonal Anti-C19orf10 antibody produced in mouse
clone 15.3.3, purified from hybridoma cell culture
Synonym(s):
IL25, IL27, IL27w, Stromal cell-derived growth factor SF20, UPF0556 protein C19orf10, myeloid-derived growth factor (MYDGF)
About This Item
biological source
mouse
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
15.3.3, monoclonal
form
buffered aqueous solution
mol wt
~17 kDa
species reactivity
bovine, canine, human
concentration
~1 mg/mL
technique(s)
flow cytometry: suitable
immunoblotting: suitable
immunofluorescence: suitable
isotype
IgG2a
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... MYDGF(56005)
General description
Immunogen
Application
- immunoblotting
- immunocytochemistry
- immunofluorescence
- flow cytometry
Biochem/physiol Actions
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class
10 - Combustible liquids
wgk_germany
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service