Skip to Content
MilliporeSigma
All Photos(4)

Key Documents

SAB4200634

Sigma-Aldrich

Monoclonal Anti-GRAMD4 antibody produced in mouse

clone GR4-1, purified from hybridoma cell culture

Synonym(s):

Death-inducing protein (DIP), GRAM domain containing 4, KIAA0767

Sign Into View Organizational & Contract Pricing

Select a Size

100 μL
$398.00

$398.00


Check Cart for Availability

Request a Bulk Order

Select a Size

Change View
100 μL
$398.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

$398.00


Check Cart for Availability

Request a Bulk Order

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

GR4-1, monoclonal

form

buffered aqueous solution

mol wt

~80 kDa

species reactivity

human, monkey

concentration

~1 mg/mL

technique(s)

flow cytometry: 2.5-5.0 μg/test using using HeLa cells.
immunoblotting: 0.5-1 μg/mL using using whole extracts of Jurkat cells.
immunocytochemistry: suitable
immunofluorescence: 2-4 μg/mL using using HeLa cells.

isotype

IgG1

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GRAMD4(23151)
mouse ... Gramd4(223752)

General description

Monoclonal Anti-GRAMD4 (mouse IgG1 isotype) is derived from the hybridoma GR4-1 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. GRAM domain containing 4 (GRAMD4) is localized in the nucleus and is mapped to human chromosome 22q13.31.

Specificity

Monoclonal Anti-GRAMD4 recognizes human and monkey GRAMD4.

Immunogen

synthetic peptide corresponding to a sequence at the N-terminal region of human GRAMD4

Application

Monoclonal Anti-GRAMD4 antibody produced in mouse has been used in:
  • immunoblotting
  • immunocytochemistry
  • flow cytometry

Biochem/physiol Actions

GRAM domain containing 4 (GRAMD4) also known as DIP (Death-inducing protein), is a novel p53- independent pro-apoptotic target of E2F Transcription Factor 1 (E2F1), which localizes to the nucleus. Upregulation of GRAMD4 by E2F1 mediates apoptosis in karyotypically normal fibroblasts and tumor cells. GRAMD4 translocation to the mitochondria is mediated by p73- induced apoptosis. It interacts with B-cell lymphoma 2 (Bcl-2), promotes BCL2 associated X, apoptosis regulator (Bax) mitochondrial relocalization and oligomerization. It induces mitochondrial membrane permeabilization mediating the release of cytochrome c and second mitochondria-derived activator of caspases (Smac). Interestingly, GRAMD4 overexpression inhibits tumor growth. Its efficacy was more pronounced over time when combined with cisplatin, leading to prolonged growth retardation.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For extended storage, freeze at -20 °Cin working aliquots. Repeated freezing and thawing,or storage in “frost-free” freezers,is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The microRNA signature of patients with sunitinib failure: regulation of UHRF1 pathways by microRNA-101 in renal cell carcinoma
Goto Y, et al.
Oncotarget, 7(37), 59070-59070 (2016)
J Stanelle et al.
Cell death and differentiation, 12(4), 347-357 (2004-11-27)
The transcription factor E2F1 does not only induce cell proliferation but also shows the strongest proapoptotic effect of all E2F family members as part of an antitumor safeguard mechanism. We have recently identified KIAA0767 as a novel p53-independent target of
K John et al.
Cell death and differentiation, 18(5), 874-886 (2010-12-04)
p73, a member of the p53 family, shares high sequence homology with p53 and shows many p53-like properties: it binds to p53-DNA target sites, transactivates p53-responsive genes and induces cell cycle arrest and apoptosis. Apart from this transcription-dependent effect, less

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service