Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

SAB4200632

Sigma-Aldrich

Monoclonal Anti-ZBBX antibody produced in mouse

clone ZBX-7, purified from hybridoma cell culture

Synonym(s):

zinc finger B-box domain containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

ZBX-7, monoclonal

form

buffered aqueous solution

species reactivity

human

concentration

~1 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using HeLa cells.
immunoblotting: suitable
immunofluorescence: 2-4 μg/mL using MCF7 cells.
immunohistochemistry: 20 μg/mL using human liver tissue sections.

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

human ... ZBBX(79740)

General description

Monoclonal Anti-ZBBX (mouse IgG2a isotype) is derived from the hybridoma Zbx-7 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a synthetic peptide. Zinc finger Bbox domain-containing protein 1 (ZBBX) is located on chromosome 3q26.1. It is a member of the B-box family.

Specificity

Monoclonal Anti- ZBBX recognizes human ZBBX.

Immunogen

synthetic peptide corresponding to an internal sequence of human ZBBX

Application

Monoclonal Anti-ZBBX antibody produced in mouse may be used in:
  • immunoblotting
  • flow cytometry
  • immunofluorescence
  • immunohistochemistry

Biochem/physiol Actions

Zinc finger Bbox domain-containing protein 1 (ZBBX) number variations is associated with attention deficit hyperactivity disorder (ADHD), a common and persistent condition, which involves developmentally atypical and impairing inattention, impulsiveness and hyperactivity. It also functions as a tumor suppressor gene in hepatocellular carcinoma (HCC).

Physical form

Supplied as a solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Storage and Stability

For extended storage, freeze at -20 °C in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilution samples should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

12 - Non Combustible Liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anath C Lionel et al.
Science translational medicine, 3(95), 95ra75-95ra75 (2011-08-13)
Attention deficit hyperactivity disorder (ADHD) is a common and persistent condition characterized by developmentally atypical and impairing inattention, hyperactivity, and impulsiveness. We identified de novo and rare copy number variations (CNVs) in 248 unrelated ADHD patients using million-feature genotyping arrays.
Lars Zender et al.
Journal of hepatology, 52(6), 921-929 (2010-04-14)
Hepatocellular carcinoma (HCC) is a deadly cancer, whose incidence is increasing worldwide. Albeit the main risk factors for HCC development have been clearly identified, such as hepatitis B and C virus infection and alcohol abuse, there is still preliminary understanding
Emma M Myers et al.
Methods (San Diego, Calif.), 73, 54-70 (2014-12-20)
Studies of the brain's transcriptome have become prominent in recent years, resulting in an accumulation of datasets with somewhat distinct attributes. These datasets, which are often analyzed only in isolation, also are often collected with divergent goals, which are reflected

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service