Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

SAB4200624

Sigma-Aldrich

Anti-EXOSC1 antibody, Mouse monoclonal

clone EX-1, purified from hybridoma cell culture

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

EX-1, monoclonal

mol wt

~21 kDa

species reactivity

mouse, human, rat

concentration

~1.0 mg/mL

technique(s)

flow cytometry: 5-10 μg/test using HeLa cells
immunoblotting: 1-2 μg/mL using whole extracts of HEK 293 cells

isotype

IgG1

UniProt accession no.

shipped in

dry ice

Storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

The exosome component 1 (EXOSC1) gene is located on human chromosome 10q24.1.
The gene EXOSC1 (exosome component 1) encodes one of the nine core subunits of the exosome complex. The protein is also referred to as Csl4 and has high affinity for RNA.

Biochem/physiol Actions

The exosome complex facilitates the splicing and degradation of mRNAs and non-coding RNAs. It is also essential for epigenetic gene regulation. It endogenously suppresses erythroid maturation. Within the complex, Exosc1 (exosome component 1) associates with Exosc6 to stabilize the complex. The binding of Exosc10 to Exosc1, Exosc6, and Exosc8 enhances the activity of the complex and positions Exosc1 for RNA binding.

Other Notes

synthetic peptide corresponding to a sequence at the internal region of human EXOSC1 conjugated to KLH

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide as a preservative.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk_germany

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eun Kyeong Oh et al.
International journal of oncology, 41(6), 2038-2046 (2012-10-02)
Host genomic alterations in addition to human papillomavirus (HPV) are needed for cervical precursor lesions to progress to invasive cancer because only a small
The RNA-Degrading Exosome Complex Is an Endogenous Suppressor of Erythroid Maturation.
McIver S
Blood, 124, 2659-2659 (2014)
Skye C McIver et al.
Blood, 124(14), 2285-2297 (2014-08-15)
Complex genetic networks control hematopoietic stem cell differentiation into progenitors that give rise to billions of erythrocytes daily. Previously, we described a role for the master regulator of erythropoiesis, GATA-1, in inducing genes encoding components of the autophagy machinery. In

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service